Keystone Cleanroom Products 1073B Tyvek™/HDPE Autoclave Stopper Bag
Autoclave Stopper Bags for Steam Sterilization. Medical Grade DuPont Tyvek and HDPE film materials yields superior seal strength for stoppers, seals and heavy equipment parts.
$1602.51 - $7745.49
Specifications
Autoclavable | Autoclavable |
---|---|
Certifications/Compliance | COC |
Closure Type | Heat Seal |
Color | White |
Disposable | Single-use |
Catalog Number | Mfr. No. | Capacity (English) | Dimensions (L x W) | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (English) | Dimensions (L x W) | Price | Quantity | |||||
19-990-259
|
Keystone Cleanroom Products
SBMGTVKHS0304 |
7 oz. | 4 x 3 in. |
Case of 1000 Each for $1,958.62
|
|
|||||
19-990-260
|
Keystone Cleanroom Products
SBMGTVKHS0306 |
10 oz. | 3 x 6 in. |
Case of 1000 Each for $2,492.80
|
|
|||||
19-990-261
|
Keystone Cleanroom Products
SBMGTVKHS0409 |
27 oz. | 9 x 4 in. |
Case of 500 Each for $1,602.51
|
|
|||||
19-990-262
|
Keystone Cleanroom Products
SBMGTVKHS0422 |
66 oz. | 22 x 4 in. |
Case of 500 Each for $2,759.89
|
|
|||||
19-990-263
|
Keystone Cleanroom Products
SBMGTVKHS0508 |
32 oz. | 8 x 5 in. |
Case of 500 Each for $1,691.55
|
|
|||||
19-990-264
|
Keystone Cleanroom Products
SBMGTVKHS0610 |
50 oz. | 10 x 6 in. |
Case of 500 Each for $2,047.65
|
|
|||||
19-990-265
|
Keystone Cleanroom Products
SBMGTVKHS0630 |
150 oz. | 30 x 6 in. |
Case of 500 Each for $4,451.42
|
|
|||||
19-990-266
|
Keystone Cleanroom Products
SBMGTVKHS0812 |
84 oz. | 12 x 8 in. |
Case of 500 Each for $3,026.97
|
|
|||||
19-990-267
|
Keystone Cleanroom Products
SBMGTVKHS1035 |
315 oz. | 35 x 10 in. |
Case of 500 Each for $6,142.97
|
|
|||||
19-990-268
|
Keystone Cleanroom Products
SBMGTVKHS1215 |
165 oz. | 15 x 12 in. |
Case of 500 Each for $4,540.45
|
|
|||||
19-990-269
|
Keystone Cleanroom Products
SBMGTVKHS1238 |
418 oz. | 38 x 12 in. |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | ||||||||||
19-990-270
|
Keystone Cleanroom Products
SBMGTVKHS1318 |
216 oz. | 18 x 13 in. |
Case of 500 Each for $5,252.69
|
|
|||||
19-990-271
|
Keystone Cleanroom Products
SBMGTVKHS1414 |
182 oz. | 14 x 14 in. |
Case of 500 Each for $4,985.60
|
|
|||||
19-990-272
|
Keystone Cleanroom Products
SBMGTVKHS1518 |
252 oz. | 18 x 15 in. |
Case of 500 Each for $5,074.63
|
|
|||||
19-990-273
|
Keystone Cleanroom Products
SBMGTVKHS1616 |
240 oz. | 16 x 16 in. |
Case of 500 Each for $5,252.69
|
|
|||||
19-990-274
|
Keystone Cleanroom Products
SBMGTVKHS1632 |
480 oz. | 32 x 16 in. |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | ||||||||||
19-990-275
|
Keystone Cleanroom Products
SBMGTVKHS1824 |
408 oz. | 24 x 18 in. |
Case of 500 Each for $6,321.02
|
|
|||||
19-990-276
|
Keystone Cleanroom Products
SBMGTVKHS2022 |
418 oz. | 22 x 20 in. |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | ||||||||||
19-990-277
|
Keystone Cleanroom Products
SBMGTVKHS2216 |
336 oz. | 16 x 22 in. |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | ||||||||||
19-990-278
|
Keystone Cleanroom Products
SBMGTVKHS2418 |
414 oz. | 18 x 24 in. |
Case of 250 Each for $3,338.57
|
|
|||||
19-990-279
|
Keystone Cleanroom Products
SBMGTVKHS2436 |
828 oz. | 36 x 24 in. |
Case of 250 Each for $5,252.69
|
|
|||||
19-990-280
|
Keystone Cleanroom Products
SBMGTVKHS2440 |
920 oz. | 40 x 24 in. |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | ||||||||||
19-990-281
|
Keystone Cleanroom Products
SBMGTVKHS2848 |
1296 oz. | 48 x 28 in. |
Case of 125 Each for $5,497.51
|
|
|||||
Description
- Designed for the Steam Sterilization of Pharmaceutical Production Equipment and Components (Stoppers and Caps)
- 1073B Tyvek™ Side Permits Steam Penetration During Sterilization
- Semi-Transpartent HDPE Film Side
- Impact Resistance
- Consistently Strong, Double Welded Factory Seals
- Exceptional Microbial Barrier
- Resists Tearing and Punctures From Sharp and Heavy Machine Parts
- Manufactured and Packaged in ISO 6 Cleanroom
- Certificates of Conformance Sent With All Shipments
Specifications
Autoclavable | |
Heat Seal | |
Single-use | |
Pharmaceutical Equipment | |
Non-sterile | |
<250°F |
COC | |
White | |
Steam Sterilization | |
Tyvek | |
Steam | |
<125°C |