Learn More
12-Bromododecylphosphonic acid, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H33714MD
Description
Employed as a intermediate for pharmaceutical.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsEmployed as a intermediate for pharmaceutical.
Notes
Store in cool, dry place in tightly closed containers. With adequate ventilation. Store away from oxidizing agent.
Specifications
12-Bromododecylphosphonic acid | |
92°C to 94°C | |
Br(CH2)12P(O)(OH)2 | |
250 mg | |
OFBOCVJPIJRGMU-UHFFFAOYSA-N | |
12-bromododecylphosphonic acid | |
45158982 | |
95% |
202920-07-8 | |
C12H26BrO3P | |
MFCD11046103 | |
12-bromododecyl phosphonic acid, phosphonic acid, 12-bromododecyl | |
OP(O)(=O)CCCCCCCCCCCCBr | |
329.22 | |
329.21 |
Chemical Identifiers
202920-07-8 | |
329.22 | |
OFBOCVJPIJRGMU-UHFFFAOYSA-N | |
45158982 | |
OP(O)(=O)CCCCCCCCCCCCBr |
C12H26BrO3P | |
MFCD11046103 | |
12-bromododecyl phosphonic acid, phosphonic acid, 12-bromododecyl | |
12-bromododecylphosphonic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only