missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

12-Bromododecylphosphonic acid, 95%, Thermo Scientific Chemicals

$312.18 - $854.60

Chemical Identifiers

CAS 202920-07-8
Molecular Formula C12H26BrO3P
Molecular Weight (g/mol) 329.22
MDL Number MFCD11046103
InChI Key OFBOCVJPIJRGMU-UHFFFAOYSA-N
Synonym 12-bromododecyl phosphonic acid, phosphonic acid, 12-bromododecyl
PubChem CID 45158982
IUPAC Name 12-bromododecylphosphonic acid
SMILES OP(O)(=O)CCCCCCCCCCCCBr
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH33714MD
View Documents
Thermo Scientific Chemicals
H33714MD
250 mg
Each for $312.18
 
AAH3371403
View Documents
Thermo Scientific Chemicals
H3371403
1 g
Each for $854.60
 
Description

Description

Employed as a intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Employed as a intermediate for pharmaceutical.

Notes
Store in cool, dry place in tightly closed containers. With adequate ventilation. Store away from oxidizing agent.
Specifications

Chemical Identifiers

202920-07-8
329.22
OFBOCVJPIJRGMU-UHFFFAOYSA-N
45158982
OP(O)(=O)CCCCCCCCCCCCBr
C12H26BrO3P
MFCD11046103
12-bromododecyl phosphonic acid, phosphonic acid, 12-bromododecyl
12-bromododecylphosphonic acid

Specifications

202920-07-8
C12H26BrO3P
MFCD11046103
12-bromododecyl phosphonic acid, phosphonic acid, 12-bromododecyl
OP(O)(=O)CCCCCCCCCCCCBr
329.22
329.21
12-Bromododecylphosphonic acid
92°C to 94°C
Br(CH2)12P(O)(OH)2
250 mg
OFBOCVJPIJRGMU-UHFFFAOYSA-N
12-bromododecylphosphonic acid
45158982
95%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only