National Safety Apparel™ 12 Cal ArcGuard™ UltraSoft Arc Flash Kit with FR Coverall and Balaclava (09)
For enhanced protection Westex™ UltraSoft is the premium choice for superior comfort, improved safety and above average durability
Supplier:  National Safety Apparel™ KIT2CV11BXL09
 View more versions of this product
Catalog No. 17001905
Explore available promotions
Description
Description
  • Arc Rating: 12 cal/cm²
  • CAT 2
  • Fabric/Material: 9 oz. Westex™ UltraSoft™
  • PureView™ faceshield technology features a greater range of visibility
  • Premium anti-fog and scratch resistant coating on faceshield
  • Action back allows for full range of motion
  • Flexible sewn-in elastic waist
  • 6 pocket style with side pass through access
  • Leg openings made to fit over work boots
  • Covered 2-way zipper
  • Made in the USA
Specifications
Specifications
12 cal/cm²
CAT 2, ASTM F1506, OSHA 1910.269, NFPA 70 E, NFPA 2112, CSA Z462
Cotton, Nylon
Zipper
None
Navy
Open
X-Large
Stitched