National Safety Apparel™ 12 Cal ArcGuard™ UltraSoft Arc Flash Kit with FR Coverall and Balaclava (09)
For enhanced protection Westex™ UltraSoft is the premium choice for superior comfort, improved safety and above average durability
Supplier: National Safety Apparel™ KIT2CV11B3X09
Description
- Arc Rating: 12 cal/cm²
- CAT 2
- Fabric/Material: 9 oz. Westex™ UltraSoft™
- PureView™ faceshield technology features a greater range of visibility
- Premium anti-fog and scratch resistant coating on faceshield
- Action back allows for full range of motion
- Flexible sewn-in elastic waist
- 6 pocket style with side pass through access
- Leg openings made to fit over work boots
- Covered 2-way zipper
- Made in the USA
Specifications
12 cal/cm² | |
CAT 2, ASTM F1506, OSHA 1910.269, NFPA 70 E, NFPA 2112, CSA Z462 | |
Cotton, Nylon | |
Zipper | |
None |
Navy | |
Open | |
3X-Large | |
Stitched |