Mopec™ #12 Sterile Scalpel Blades
High-quality, sterile blades used for anatomic pathology or autopsy.
Supplier: Mopec™ AJ205
Description
- Individually wrapped
- Fits #3, #3L and #5 scalpel handles
Specifications
Stainless Steel | |
# 12 | |
Sterile | |
No |
Scalpel | |
Single-use | |
100 Blades/Pk. |