Mopec™ #12 Sterile Scalpel Blades
High-quality, sterile blades used for anatomic pathology or autopsy.
Supplier:  Mopec™ AJ172
 View more versions of this product
Catalog No. 22-445-600
Explore available promotions
Description
Description
  • Individually wrapped
  • Fits #3, #3L and #5 scalpel handles
Specifications
Specifications
Carbon Steel
# 12
Sterile
No
Scalpel
Single-use
100 Blades/Pk.
Product Suggestions
Product Suggestions