Justrite™ 15 Gallon ChemCor™ Under Fume Hood Corrosives/Acids Safety Cabinet With Self-Closing Doors
Increase workplace safety and improve cleaniliness with the proper safety and storage equipment. The Justrite™ 15 Gallon ChemCor™ Under Fume Hood Corrosives/Acids Safety Cabinet With Self-Closing Doors provides superior corrosion protection as well as fire protection in a single cabinet.
Supplier: Justrite™ 8825222
Description
- Expand storage, add new convenience to existing cabinets, or customize your new cabinet with additional shelves to meet specific workflow needs
- Galvanized steel shelves hold up to 250 lbs and adjust on 3 in. increments for versatile storage
- Shelves have the ChemCor thermoplastic coating and provide superior protection against corrosion from drips and off-gassing from harsh chemicals
- Patented SpillSlope galvanized steel shelves direct spills to back and bottom of a leakproof sump
Specifications
Blue | |
Right Door | |
15 gal. | |
Self-close |