missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1H-Benzotriazole, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 183885000

 View more versions of this product

Catalog No. AC183885000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
Specifications

Specifications

1H-Benzotriazole
95-14-7
100.0
Beige-Yellow to White
170°C
99%
C6H5N3
500 g
16,21
1h-benzotriazole, benzotriazole, 1,2,3-benzotriazole, 1h-benzo d 1,2,3 triazole, azimidobenzene, 1h-1,2,3-benzotriazole, aziminobenzene, benztriazole, benzene azimide, benzisotriazole
QRUDEWIWKLJBPS-UHFFFAOYSA-N
2H-benzotriazole
7220
119.13
Powder, Granules, Crystals, Needles or Flakes
98%
98.5
94.0°C to 99.0°C
204.0°C (15.0 mmHg)
Authentic
Plastic bottle
MFCD00005699
26, 38
15, 1111
Solubility in water: 25g/L in water (20°C). Other solubilities: soluble in alcohol,benzene,toluene,chloroform,and dmf,soluble in ethanol,methanol and ethyleneglycol
C1=CC2=NNN=C2C=C1
119.13
CHEBI:75331
99%

Chemical Identifiers

95-14-7
119.13
QRUDEWIWKLJBPS-UHFFFAOYSA-N
7220
2H-benzotriazole
C6H5N3
MFCD00005699
1h-benzotriazole, benzotriazole, 1,2,3-benzotriazole, 1h-benzo d 1,2,3 triazole, azimidobenzene, 1h-1,2,3-benzotriazole, aziminobenzene, benztriazole, benzene azimide, benzisotriazole
CHEBI:75331
C1=CC2=NNN=C2C=C1
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes serious eye irritation.
Toxic to aquatic life with long lasting effects.

GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning

EINECSNumber : 202-394-1

RTECSNumber : DM1225000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only