missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1H-Pyrazole, 98%, Thermo Scientific Chemicals

1H-Pyrazole, 98%, C3H4N2, CAS Number-288-13-1, 1-h-pyrazole, 1h-pyrazol, pyrazole, unii-3qd5kjz7zj, hpz, pyrazol, diazole, pyrazol#1, 1,2-diazole, 3qd5kjz7zj, 100g, 103775, 186 deg.C to 188 deg.C, CHEBI:17241, 206-017-1, 68.08, H302-H315-H319-H335

Supplier:  Thermo Scientific Chemicals A1418622

 View more versions of this product

Catalog No. AAA1418622


Explore available promotions

Description

Description

1H-Pyrazole is used as a ligand in the preparation of organometallic compounds. The derivatives of Pyrazole are used in medicine, for their analgesic, anti-inflammatory, antipyretic, antiarrhythmic, tranquilizing, muscle relaxing, psychoanaleptic, anticonvulsant, monoamineoxidase inhibiting, antidiabetic and antibacterial activities.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1H-Pyrazole is used as a ligand in the preparation of organometallic compounds. The derivatives of Pyrazole are used in medicine, for their analgesic, anti-inflammatory, antipyretic, antiarrhythmic, tranquilizing, muscle relaxing, psychoanaleptic, anticonvulsant, monoamineoxidase inhibiting, antidiabetic and antibacterial activities.

Solubility
Soluble in water.

Notes
Hygroscopic. Store in cool, dry place in tightly closed containers. Store away from oxidizing agent and water/moisture.
Specifications

Specifications

1H-Pyrazole
66°C to 70°C
Unpleasant
MFCD00005234
103775
14,7960
Soluble in water.
C1=CNN=C1
68.079
CHEBI:17241
98%
288-13-1
186°C to 188°C
C3H4N2
100 g
Hygroscopic
pyrazole, 1,2-diazole, 1h-pyrazol, pyrazol, unii-3qd5kjz7zj, 3qd5kjz7zj, diazole, hpz, 1-h-pyrazole, pyrazol#1
WTKZEGDFNFYCGP-UHFFFAOYSA-N
1H-pyrazole
1048
68.08

Chemical Identifiers

288-13-1
68.079
WTKZEGDFNFYCGP-UHFFFAOYSA-N
1048
1H-pyrazole
C3H4N2
MFCD00005234
pyrazole, 1,2-diazole, 1h-pyrazol, pyrazol, unii-3qd5kjz7zj, 3qd5kjz7zj, diazole, hpz, 1-h-pyrazole, pyrazol#1
CHEBI:17241
C1=CNN=C1
Safety and Handling

Safety and Handling

GHS H Statement
H361-H302-H315-H319-H335
Suspected of damaging fertility or the unborn child.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

EINECSNumber : 206-017-1

RTECSNumber : UQ4900000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Argon

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only