Learn More
1H-Pyrazole, 98%
1H-Pyrazole, 98%, C3H4N2, CAS Number-288-13-1, 1-h-pyrazole, 1h-pyrazol, pyrazole, unii-3qd5kjz7zj, hpz, pyrazol, diazole, pyrazol#1, 1,2-diazole, 3qd5kjz7zj, 100g, 103775, 186 deg.C to 188 deg.C, CHEBI:17241, 206-017-1, 68.08, H302-H315-H319-H335 | CAS: 288-13-1 | C3H4N2 | 68.079 g/mol
$107.68 - $301.86
Chemical Identifiers
| CAS | 288-13-1 |
|---|---|
| Molecular Formula | C3H4N2 |
| Molecular Weight (g/mol) | 68.079 |
| MDL Number | MFCD00005234 |
| InChI Key | WTKZEGDFNFYCGP-UHFFFAOYSA-N |
| Synonym | pyrazole, 1,2-diazole, 1h-pyrazol, pyrazol, unii-3qd5kjz7zj, 3qd5kjz7zj, diazole, hpz, 1-h-pyrazole, pyrazol#1 |
| PubChem CID | 1048 |
| ChEBI | CHEBI:17241 |
| IUPAC Name | 1H-pyrazole |
| SMILES | C1=CNN=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1418614
|
Thermo Scientific Chemicals
A1418614 |
25 g |
Each for $107.68
|
|
|||||
|
AAA1418622
|
Thermo Scientific Chemicals
A1418622 |
100 g |
Each for $301.86
|
|
|||||
Description
1H-Pyrazole is used as a ligand in the preparation of organometallic compounds. The derivatives of Pyrazole are used in medicine, for their analgesic, anti-inflammatory, antipyretic, antiarrhythmic, tranquilizing, muscle relaxing, psychoanaleptic, anticonvulsant, monoamineoxidase inhibiting, antidiabetic and antibacterial activities.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1H-Pyrazole is used as a ligand in the preparation of organometallic compounds. The derivatives of Pyrazole are used in medicine, for their analgesic, anti-inflammatory, antipyretic, antiarrhythmic, tranquilizing, muscle relaxing, psychoanaleptic, anticonvulsant, monoamineoxidase inhibiting, antidiabetic and antibacterial activities.
Solubility
Soluble in water.
Notes
Hygroscopic. Store in cool, dry place in tightly closed containers. Store away from oxidizing agent and water/moisture.
Chemical Identifiers
| 288-13-1 | |
| 68.079 | |
| WTKZEGDFNFYCGP-UHFFFAOYSA-N | |
| 1048 | |
| 1H-pyrazole |
| C3H4N2 | |
| MFCD00005234 | |
| pyrazole, 1,2-diazole, 1h-pyrazol, pyrazol, unii-3qd5kjz7zj, 3qd5kjz7zj, diazole, hpz, 1-h-pyrazole, pyrazol#1 | |
| CHEBI:17241 | |
| C1=CNN=C1 |
Specifications
| 288-13-1 | |
| 186°C to 188°C | |
| C3H4N2 | |
| 25 g | |
| Hygroscopic | |
| pyrazole, 1,2-diazole, 1h-pyrazol, pyrazol, unii-3qd5kjz7zj, 3qd5kjz7zj, diazole, hpz, 1-h-pyrazole, pyrazol#1 | |
| WTKZEGDFNFYCGP-UHFFFAOYSA-N | |
| 1H-pyrazole | |
| 1048 | |
| 68.08 | |
| 1H-Pyrazole |
| 66°C to 70°C | |
| Unpleasant | |
| MFCD00005234 | |
| 103775 | |
| 14,7960 | |
| Soluble in water. | |
| C1=CNN=C1 | |
| 68.079 | |
| CHEBI:17241 | |
| 98% |
Safety and Handling
GHS H Statement
H361-H302-H315-H319-H335
Suspected of damaging fertility or the unborn child.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
EINECSNumber : 206-017-1
RTECSNumber : UQ4900000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only