missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1H-Tetrazole, 0.45M in acetonitrile, Thermo Scientific Chemicals

Coupling reagent for preparation of polynucleotides

$567.95 - $1137.71

Chemical Identifiers

CAS 288-94-8
Molecular Formula CH2N4
Molecular Weight (g/mol) 70.055
MDL Number MFCD00005247
InChI Key KJUGUADJHNHALS-UHFFFAOYSA-N
PubChem CID 67519
ChEBI CHEBI:33193
IUPAC Name 2H-tetrazole
SMILES C1=NNN=N1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ61034AH
View Documents
Thermo Scientific Chemicals
J61034AH
200 mL
Each for $567.95
 
AAJ61034AO
View Documents
Thermo Scientific Chemicals
J61034AO
450 mL
Each for $1,137.71
 
Description

Description

1H-Tetrazole is used as a bioisostere for the carboxylate group. It is also used as coupling reagent for preparation of polynucleotides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1H-Tetrazole is used as a bioisostere for the carboxylate group. It is also used as coupling reagent for preparation of polynucleotides.

Solubility
Miscible with water.

Notes
Incompatible with reducing agents and alkali metals.
Specifications

Chemical Identifiers

288-94-8
70.055
KJUGUADJHNHALS-UHFFFAOYSA-N
CHEBI:33193
C1=NNN=N1
CH2N4
MFCD00005247
67519
2H-tetrazole

Specifications

288-94-8
CH2N4
200 mL
105799
1h-tetrazole, tetrazole, tetraazacyclopentadiene, 1-h-tetrazole, 1h-1,2,3,4-tetrazole, 1 h-tetrazole, 1h-tetrazole solution, 2h-1,2,3,4-tetrazole, racemic tetrazole, 1h-tetraazole
KJUGUADJHNHALS-UHFFFAOYSA-N
2H-tetrazole
67519
70.05
Liquid
0.798
MFCD00005247
UN1648
14,2786
Miscible with water.
C1=NNN=N1
70.055
CHEBI:33193
0.45M in acetonitrile
1H-Tetrazole
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H312-H332-H319
Highly flammable liquid and vapor.
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye irritation.

P210-P233-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P363-P370+P378q-P501c

H225-H302+H312+H332-H319

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ACETONITRILE

EINECSNumber : 206-023-4

RTECSNumber : UW7370000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only