missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(1R,2R)-(+)-1,2-Diaminocyclohexane L-tartrate, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H6017209

 View more versions of this product

Catalog No. AAH6017209


Explore available promotions

Description

Description

(1R,2R)-(+)-1,2-Diaminocyclohexane L-tartrate is a useful chemical for the synthesis of chiral ligands.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(1R,2R)-(+)-1,2-Diaminocyclohexane L-tartrate is a useful chemical for the synthesis of chiral ligands.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Specifications

(1R,2R)-(+)-1,2-Diaminocyclohexane L-tartrate
273°C (decomposition)
MFCD00191979
1r,2r-+-1,2-diaminocyclohexane l-tartrate, 1r,2r-cyclohexane-1,2-diamine 2r,3r-2,3-dihydroxysuccinate, 1r,2r-+-1,2-cyclohexanediamine l-tartrate, 1r,2r---cyclohexane-1,2-diamine l-tartrate salt, 1r-trans-1,2-diaminocyclohexane l-tartrate, 1r,2r-+-cyclohexane-1,2-diamine l-tartrate, 1r,2r-1,2-diaminocyclohexane l-tartrate, 1r,2r-cyclohexane-1,2-diamine; l +-tartaric acid, pubchem17364, ksc221k4n
+12° (c=4 in Water)
(1R,2R)-cyclohexane-1,2-diamine;(2R,3R)-2,3-dihydroxybutanedioic acid
11448443
99%
39961-95-0
C10H20N2O6
10 g
GDOTUTAQOJUZOF-ZXZVGZDWSA-N
C1CCC(C(C1)N)N.C(C(C(=O)O)O)(C(=O)O)O
264.278
264.28

Chemical Identifiers

39961-95-0
264.278
GDOTUTAQOJUZOF-ZXZVGZDWSA-N
11448443
C1CCC(C(C1)N)N.C(C(C(=O)O)O)(C(=O)O)O
C10H20N2O6
MFCD00191979
1r,2r-+-1,2-diaminocyclohexane l-tartrate, 1r,2r-cyclohexane-1,2-diamine 2r,3r-2,3-dihydroxysuccinate, 1r,2r-+-1,2-cyclohexanediamine l-tartrate, 1r,2r---cyclohexane-1,2-diamine l-tartrate salt, 1r-trans-1,2-diaminocyclohexane l-tartrate, 1r,2r-+-cyclohexane-1,2-diamine l-tartrate, 1r,2r-1,2-diaminocyclohexane l-tartrate, 1r,2r-cyclohexane-1,2-diamine; l +-tartaric acid, pubchem17364, ksc221k4n
(1R,2R)-cyclohexane-1,2-diamine;(2R,3R)-2,3-dihydroxybutanedioic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only