Learn More
(1R,2R)-(+)-1,2-Diaminocyclohexane L-tartrate, 98%, Thermo Scientific Chemicals
$61.27 - $259.12
Chemical Identifiers
CAS | 39961-95-0 |
---|---|
Molecular Formula | C10H20N2O6 |
Molecular Weight (g/mol) | 264.278 |
MDL Number | MFCD00191979 |
InChI Key | GDOTUTAQOJUZOF-ZXZVGZDWSA-N |
Synonym | 1r,2r-+-1,2-diaminocyclohexane l-tartrate, 1r,2r-cyclohexane-1,2-diamine 2r,3r-2,3-dihydroxysuccinate, 1r,2r-+-1,2-cyclohexanediamine l-tartrate, 1r,2r---cyclohexane-1,2-diamine l-tartrate salt, 1r-trans-1,2-diaminocyclohexane l-tartrate, 1r,2r-+-cyclohexane-1,2-diamine l-tartrate, 1r,2r-1,2-diaminocyclohexane l-tartrate, 1r,2r-cyclohexane-1,2-diamine; l +-tartaric acid, pubchem17364, ksc221k4n |
PubChem CID | 11448443 |
IUPAC Name | (1R,2R)-cyclohexane-1,2-diamine;(2R,3R)-2,3-dihydroxybutanedioic acid |
SMILES | C1CCC(C(C1)N)N.C(C(C(=O)O)O)(C(=O)O)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH6017204
|
Thermo Scientific Chemicals
H6017204 |
2 g |
Each for $61.27
|
|
|||||
AAH6017209
|
Thermo Scientific Chemicals
H6017209 |
10 g |
Each for $259.12
|
|
|||||
Description
(1R,2R)-(+)-1,2-Diaminocyclohexane L-tartrate is a useful chemical for the synthesis of chiral ligands.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications(1R,2R)-(+)-1,2-Diaminocyclohexane L-tartrate is a useful chemical for the synthesis of chiral ligands.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Chemical Identifiers
39961-95-0 | |
264.278 | |
GDOTUTAQOJUZOF-ZXZVGZDWSA-N | |
11448443 | |
C1CCC(C(C1)N)N.C(C(C(=O)O)O)(C(=O)O)O |
C10H20N2O6 | |
MFCD00191979 | |
1r,2r-+-1,2-diaminocyclohexane l-tartrate, 1r,2r-cyclohexane-1,2-diamine 2r,3r-2,3-dihydroxysuccinate, 1r,2r-+-1,2-cyclohexanediamine l-tartrate, 1r,2r---cyclohexane-1,2-diamine l-tartrate salt, 1r-trans-1,2-diaminocyclohexane l-tartrate, 1r,2r-+-cyclohexane-1,2-diamine l-tartrate, 1r,2r-1,2-diaminocyclohexane l-tartrate, 1r,2r-cyclohexane-1,2-diamine; l +-tartaric acid, pubchem17364, ksc221k4n | |
(1R,2R)-cyclohexane-1,2-diamine;(2R,3R)-2,3-dihydroxybutanedioic acid |
Specifications
39961-95-0 | |
C10H20N2O6 | |
2 g | |
GDOTUTAQOJUZOF-ZXZVGZDWSA-N | |
C1CCC(C(C1)N)N.C(C(C(=O)O)O)(C(=O)O)O | |
264.278 | |
264.28 | |
(1R,2R)-(+)-1,2-Diaminocyclohexane L-tartrate |
273°C (decomposition) | |
MFCD00191979 | |
1r,2r-+-1,2-diaminocyclohexane l-tartrate, 1r,2r-cyclohexane-1,2-diamine 2r,3r-2,3-dihydroxysuccinate, 1r,2r-+-1,2-cyclohexanediamine l-tartrate, 1r,2r---cyclohexane-1,2-diamine l-tartrate salt, 1r-trans-1,2-diaminocyclohexane l-tartrate, 1r,2r-+-cyclohexane-1,2-diamine l-tartrate, 1r,2r-1,2-diaminocyclohexane l-tartrate, 1r,2r-cyclohexane-1,2-diamine; l +-tartaric acid, pubchem17364, ksc221k4n | |
+12° (c=4 in Water) | |
(1R,2R)-cyclohexane-1,2-diamine;(2R,3R)-2,3-dihydroxybutanedioic acid | |
11448443 | |
99% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only