missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
(1R,3S)-(+)-Camphoric acid, 98+%, Thermo Scientific Chemicals
$59.36 - $551.51
Chemical Identifiers
CAS | 124-83-4 |
---|---|
Molecular Formula | C10H14O4-2 |
Molecular Weight (g/mol) | 198.218 |
MDL Number | MFCD00001375 |
InChI Key | LSPHULWDVZXLIL-LDWIPMOCSA-L |
Synonym | d-+-camphoric acid, 1r,3s-1,2,2-trimethylcyclopentane-1,3-dicarboxylic acid, d-camphoric acid, 1r,3s-+-camphoric acid, unii-w77rm1csd5, camphoric acid, 1,3-cyclopentanedicarboxylic acid, 1,2,2-trimethyl-, 1r,3s, w77rm1csd5, 1,2,2-trimethyl-1,3-cyclopentanedicarboxylic acid, 1r-cis, 1r,3s-camphoric acid |
PubChem CID | 6918944 |
IUPAC Name | (1R,3S)-1,2,2-trimethylcyclopentane-1,3-dicarboxylate |
SMILES | CC1(C(CCC1(C)C(=O)[O-])C(=O)[O-])C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2310614
|
Thermo Scientific Chemicals
B2310614 |
25 g |
Each for $59.36
|
|
|||||
AAB2310622
|
Thermo Scientific Chemicals
B2310622 |
100 g |
Each for $166.18
|
|
|||||
AAB2310636
|
Thermo Scientific Chemicals
B2310636 |
500 g |
Each for $551.51
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
124-83-4 | |
198.218 | |
LSPHULWDVZXLIL-LDWIPMOCSA-L | |
6918944 | |
CC1(C(CCC1(C)C(=O)[O-])C(=O)[O-])C |
C10H14O4-2 | |
MFCD00001375 | |
d-+-camphoric acid, 1r,3s-1,2,2-trimethylcyclopentane-1,3-dicarboxylic acid, d-camphoric acid, 1r,3s-+-camphoric acid, unii-w77rm1csd5, camphoric acid, 1,3-cyclopentanedicarboxylic acid, 1,2,2-trimethyl-, 1r,3s, w77rm1csd5, 1,2,2-trimethyl-1,3-cyclopentanedicarboxylic acid, 1r-cis, 1r,3s-camphoric acid | |
(1R,3S)-1,2,2-trimethylcyclopentane-1,3-dicarboxylate |
Specifications
183°C to 188°C | |
C10H14O4-2 | |
2050204 | |
d-+-camphoric acid, 1r,3s-1,2,2-trimethylcyclopentane-1,3-dicarboxylic acid, d-camphoric acid, 1r,3s-+-camphoric acid, unii-w77rm1csd5, camphoric acid, 1,3-cyclopentanedicarboxylic acid, 1,2,2-trimethyl-, 1r,3s, w77rm1csd5, 1,2,2-trimethyl-1,3-cyclopentanedicarboxylic acid, 1r-cis, 1r,3s-camphoric acid | |
CC1(C(CCC1(C)C(=O)[O-])C(=O)[O-])C | |
198.218 | |
200.23 | |
25 g | |
(1R,3S)-(+)-Camphoric acid |
124-83-4 | |
MFCD00001375 | |
141733 | |
LSPHULWDVZXLIL-LDWIPMOCSA-L | |
(1R,3S)-1,2,2-trimethylcyclopentane-1,3-dicarboxylate | |
6918944 | |
≥98% | |
+46° (c=1 in Methanol) |
Safety and Handling
EINECSNumber : 204-715-0
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only