missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(1S,2S)-2-(Diphenylphosphino)-1,2-diphenylethylamine, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 434092500

 View more versions of this product

Catalog No. AC434092500


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

(1S, 2S)-2-(Diphenylphosphino)-1, 2-diphenylethylamine
96.0
87.0°C to 93.0°C
Authentic
Glass bottle
MFCD11045444
1s,2s-2-diphenylphosphino-1,2-diphenylethanamine, 1s,2s-2-diphenylphosphino-1,2-diphenylethylamine, 1s,2s-2-amino-1,2-diphenylethyl diphenylphosphane, 1s,2s-1,2-diphenyl-2-aminoethyl diphenylphosphine, 1s,2s-2-diphenylphosphanyl-1,2-diphenylethan-1-amine, s,s-+-2-diphenylphosphino-1,2-diphenylethylamine, 1s,2s-2-diphenylphosphino-1,2-diphenylethylamine, kanata purity
N[C@H]([C@@H](P(C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1
381.46
381.45
Crystalline Powder
1091606-67-5
100.0
White to Yellow
97%
C26H24NP
250 mg
MIPGTOGPLXZBQX-QYRYXDJWNA-N
(1S,2S)-2-diphenylphosphanyl-1,2-diphenylethanamine
46177587
97%

Chemical Identifiers

1091606-67-5
381.46
MIPGTOGPLXZBQX-QYRYXDJWNA-N
46177587
N[C@H]([C@@H](P(C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1
C26H24NP
MFCD11045444
1s,2s-2-diphenylphosphino-1,2-diphenylethanamine, 1s,2s-2-diphenylphosphino-1,2-diphenylethylamine, 1s,2s-2-amino-1,2-diphenylethyl diphenylphosphane, 1s,2s-1,2-diphenyl-2-aminoethyl diphenylphosphine, 1s,2s-2-diphenylphosphanyl-1,2-diphenylethan-1-amine, s,s-+-2-diphenylphosphino-1,2-diphenylethylamine, 1s,2s-2-diphenylphosphino-1,2-diphenylethylamine, kanata purity
(1S,2S)-2-diphenylphosphanyl-1,2-diphenylethanamine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only