missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(1S,2S)-2-(Diphenylphosphino)-1,2-diphenylethylamine, 97%, Thermo Scientific Chemicals

$302.98 - $302.98

Chemical Identifiers

CAS 1091606-67-5
Molecular Formula C26H24NP
Molecular Weight (g/mol) 381.46
MDL Number MFCD11045444
InChI Key MIPGTOGPLXZBQX-QYRYXDJWNA-N
Synonym 1s,2s-2-diphenylphosphino-1,2-diphenylethanamine, 1s,2s-2-diphenylphosphino-1,2-diphenylethylamine, 1s,2s-2-amino-1,2-diphenylethyl diphenylphosphane, 1s,2s-1,2-diphenyl-2-aminoethyl diphenylphosphine, 1s,2s-2-diphenylphosphanyl-1,2-diphenylethan-1-amine, s,s-+-2-diphenylphosphino-1,2-diphenylethylamine, 1s,2s-2-diphenylphosphino-1,2-diphenylethylamine, kanata purity
PubChem CID 46177587
IUPAC Name (1S,2S)-2-diphenylphosphanyl-1,2-diphenylethanamine
SMILES N[C@H]([C@@H](P(C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC434092500
SDS View Documents
Thermo Scientific Chemicals
434092500
250 mg Glass bottle
Each for $302.98
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1091606-67-5
381.46
MIPGTOGPLXZBQX-QYRYXDJWNA-N
46177587
N[C@H]([C@@H](P(C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1
C26H24NP
MFCD11045444
1s,2s-2-diphenylphosphino-1,2-diphenylethanamine, 1s,2s-2-diphenylphosphino-1,2-diphenylethylamine, 1s,2s-2-amino-1,2-diphenylethyl diphenylphosphane, 1s,2s-1,2-diphenyl-2-aminoethyl diphenylphosphine, 1s,2s-2-diphenylphosphanyl-1,2-diphenylethan-1-amine, s,s-+-2-diphenylphosphino-1,2-diphenylethylamine, 1s,2s-2-diphenylphosphino-1,2-diphenylethylamine, kanata purity
(1S,2S)-2-diphenylphosphanyl-1,2-diphenylethanamine

Specifications

1091606-67-5
100.0
White to Yellow
97%
C26H24NP
250 mg
MIPGTOGPLXZBQX-QYRYXDJWNA-N
(1S,2S)-2-diphenylphosphanyl-1,2-diphenylethanamine
46177587
97%
(1S, 2S)-2-(Diphenylphosphino)-1, 2-diphenylethylamine
96.0
87.0°C to 93.0°C
Authentic
Glass bottle
MFCD11045444
1s,2s-2-diphenylphosphino-1,2-diphenylethanamine, 1s,2s-2-diphenylphosphino-1,2-diphenylethylamine, 1s,2s-2-amino-1,2-diphenylethyl diphenylphosphane, 1s,2s-1,2-diphenyl-2-aminoethyl diphenylphosphine, 1s,2s-2-diphenylphosphanyl-1,2-diphenylethan-1-amine, s,s-+-2-diphenylphosphino-1,2-diphenylethylamine, 1s,2s-2-diphenylphosphino-1,2-diphenylethylamine, kanata purity
N[C@H]([C@@H](P(C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1
381.46
381.45
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only