missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2,2,3,3-Tetrafluoro-1,4-butanediol 95.0+%, TCI America™
$39.81 - $524.24
Chemical Identifiers
CAS | 425-61-6 |
---|---|
Molecular Formula | C4H6F4O2 |
Molecular Weight (g/mol) | 162.08 |
MDL Number | MFCD00042375 |
InChI Key | CDZXJJOGDCLNKX-UHFFFAOYSA-N |
Synonym | 2,2,3,3-tetrafluoro-1,4-butanediol, 1,4-dihydroxy-2,2,3,3-tetrafluorobutane, acmc-1aqx7, 1,4-butanediol,2,2,3,3-tetrafluoro, 2,2,3,3-tetrakis fluoranyl butane-1,4-diol |
PubChem CID | 136270 |
IUPAC Name | 2,2,3,3-tetrafluorobutane-1,4-diol |
SMILES | OCC(F)(F)C(F)(F)CO |
Chemical Identifiers
425-61-6 | |
162.08 | |
CDZXJJOGDCLNKX-UHFFFAOYSA-N | |
136270 | |
OCC(F)(F)C(F)(F)CO |
C4H6F4O2 | |
MFCD00042375 | |
2,2,3,3-tetrafluoro-1,4-butanediol, 1,4-dihydroxy-2,2,3,3-tetrafluorobutane, acmc-1aqx7, 1,4-butanediol,2,2,3,3-tetrafluoro, 2,2,3,3-tetrakis fluoranyl butane-1,4-diol | |
2,2,3,3-tetrafluorobutane-1,4-diol |
Specifications
425-61-6 | |
White | |
C4H6F4O2 | |
1 g | |
CDZXJJOGDCLNKX-UHFFFAOYSA-N | |
2,2,3,3-tetrafluorobutane-1,4-diol | |
136270 | |
≥95.0% (GC) | |
2,2,3,3-Tetrafluoro-1,4-butanediol |
80°C | |
112°C | |
MFCD00042375 | |
2,2,3,3-tetrafluoro-1,4-butanediol, 1,4-dihydroxy-2,2,3,3-tetrafluorobutane, acmc-1aqx7, 1,4-butanediol,2,2,3,3-tetrafluoro, 2,2,3,3-tetrakis fluoranyl butane-1,4-diol | |
OCC(F)(F)C(F)(F)CO | |
162.08 | |
162.08 | |
Crystalline Powder |
Safety and Handling
TSCA : No