missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2,2,3,3-Tetrafluoro-1,4-butanediol 95.0+%, TCI America™
Supplier: TCI America T22961G
Specifications
2,2,3,3-Tetrafluoro-1,4-butanediol | |
80°C | |
112°C | |
MFCD00042375 | |
2,2,3,3-tetrafluoro-1,4-butanediol, 1,4-dihydroxy-2,2,3,3-tetrafluorobutane, acmc-1aqx7, 1,4-butanediol,2,2,3,3-tetrafluoro, 2,2,3,3-tetrakis fluoranyl butane-1,4-diol | |
OCC(F)(F)C(F)(F)CO | |
162.08 | |
162.08 | |
Crystalline Powder |
425-61-6 | |
White | |
C4H6F4O2 | |
1 g | |
CDZXJJOGDCLNKX-UHFFFAOYSA-N | |
2,2,3,3-tetrafluorobutane-1,4-diol | |
136270 | |
≥95.0% (GC) |
Chemical Identifiers
425-61-6 | |
162.08 | |
CDZXJJOGDCLNKX-UHFFFAOYSA-N | |
136270 | |
OCC(F)(F)C(F)(F)CO |
C4H6F4O2 | |
MFCD00042375 | |
2,2,3,3-tetrafluoro-1,4-butanediol, 1,4-dihydroxy-2,2,3,3-tetrafluorobutane, acmc-1aqx7, 1,4-butanediol,2,2,3,3-tetrafluoro, 2,2,3,3-tetrakis fluoranyl butane-1,4-diol | |
2,2,3,3-tetrafluorobutane-1,4-diol |
Safety and Handling
TSCA : No