missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,2,4-Trimethyl-1,3-pentanediolmono(2-methylpropanoate), 98+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 422010025

 View more versions of this product

Catalog No. AC422010025


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2,2,4-Trimethyl-1,3-pentanediolmono(2-methylpropanoate)
98.0
0.9500g/mL
114°C
98% min. (sum two isomers) (GC)
C12H24O3
MFCD00148967
0.95
DAFHKNAQFPVRKR-UHFFFAOYSA-N
(3-hydroxy-2,2,4-trimethylpentyl) 2-methylpropanoate
6490
216.32
Liquid
25265-77-4
100.0
244.0°C
Authentic
Glass bottle
1.4400 to 1.4420
2.5 kg
texanol, 2,2,4-trimethyl-1,3-pentanediol monoisobutyrate, chissocizer cs 12, propanoic acid, 2-methyl-, 3-hydroxy-2,2,4-trimethylpentyl ester, 2,2,4-trimethylpentane-1,3-diol monoisobutyrate, 3-hydroxy-2,2,4-trimethylpentyl isobutyrate, ccris 8966, isobutyraldehyde tishchenko trimer, propanoic acid, 2-methyl-, monoester with 2,2,4-trimethyl-1,3-pentanediol, 2,2,4-trimethyl-1,3-pentanediolmono 2-methylpropanoate
CC(C)C(C(C)(C)COC(=O)C(C)C)O
216.32
16 mPa.s (20°C)
98+%

Chemical Identifiers

25265-77-4
216.32
DAFHKNAQFPVRKR-UHFFFAOYSA-N
6490
CC(C)C(C(C)(C)COC(=O)C(C)C)O
C12H24O3
MFCD00148967
texanol, 2,2,4-trimethyl-1,3-pentanediol monoisobutyrate, chissocizer cs 12, propanoic acid, 2-methyl-, 3-hydroxy-2,2,4-trimethylpentyl ester, 2,2,4-trimethylpentane-1,3-diol monoisobutyrate, 3-hydroxy-2,2,4-trimethylpentyl isobutyrate, ccris 8966, isobutyraldehyde tishchenko trimer, propanoic acid, 2-methyl-, monoester with 2,2,4-trimethyl-1,3-pentanediol, 2,2,4-trimethyl-1,3-pentanediolmono 2-methylpropanoate
(3-hydroxy-2,2,4-trimethylpentyl) 2-methylpropanoate
Safety and Handling

Safety and Handling

EINECSNumber : 246-771-9

RTECSNumber : UF6000000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only