missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,2,4-Trimethyl-1,3-pentanediolmono(2-methylpropanoate), 98+%, Thermo Scientific Chemicals

$171.75 - $171.75

Chemical Identifiers

CAS 25265-77-4
Molecular Formula C12H24O3
Molecular Weight (g/mol) 216.32
MDL Number MFCD00148967
InChI Key DAFHKNAQFPVRKR-UHFFFAOYSA-N
Synonym texanol, 2,2,4-trimethyl-1,3-pentanediol monoisobutyrate, chissocizer cs 12, propanoic acid, 2-methyl-, 3-hydroxy-2,2,4-trimethylpentyl ester, 2,2,4-trimethylpentane-1,3-diol monoisobutyrate, 3-hydroxy-2,2,4-trimethylpentyl isobutyrate, ccris 8966, isobutyraldehyde tishchenko trimer, propanoic acid, 2-methyl-, monoester with 2,2,4-trimethyl-1,3-pentanediol, 2,2,4-trimethyl-1,3-pentanediolmono 2-methylpropanoate
PubChem CID 6490
IUPAC Name (3-hydroxy-2,2,4-trimethylpentyl) 2-methylpropanoate
SMILES CC(C)C(C(C)(C)COC(=O)C(C)C)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC422010025
SDS View Documents
Thermo Scientific Chemicals
422010025
2.5 kg Glass bottle
Each for $171.75
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

25265-77-4
216.32
DAFHKNAQFPVRKR-UHFFFAOYSA-N
6490
CC(C)C(C(C)(C)COC(=O)C(C)C)O
C12H24O3
MFCD00148967
texanol, 2,2,4-trimethyl-1,3-pentanediol monoisobutyrate, chissocizer cs 12, propanoic acid, 2-methyl-, 3-hydroxy-2,2,4-trimethylpentyl ester, 2,2,4-trimethylpentane-1,3-diol monoisobutyrate, 3-hydroxy-2,2,4-trimethylpentyl isobutyrate, ccris 8966, isobutyraldehyde tishchenko trimer, propanoic acid, 2-methyl-, monoester with 2,2,4-trimethyl-1,3-pentanediol, 2,2,4-trimethyl-1,3-pentanediolmono 2-methylpropanoate
(3-hydroxy-2,2,4-trimethylpentyl) 2-methylpropanoate

Specifications

25265-77-4
100.0
244.0°C
Authentic
Glass bottle
1.4400 to 1.4420
2.5 kg
texanol, 2,2,4-trimethyl-1,3-pentanediol monoisobutyrate, chissocizer cs 12, propanoic acid, 2-methyl-, 3-hydroxy-2,2,4-trimethylpentyl ester, 2,2,4-trimethylpentane-1,3-diol monoisobutyrate, 3-hydroxy-2,2,4-trimethylpentyl isobutyrate, ccris 8966, isobutyraldehyde tishchenko trimer, propanoic acid, 2-methyl-, monoester with 2,2,4-trimethyl-1,3-pentanediol, 2,2,4-trimethyl-1,3-pentanediolmono 2-methylpropanoate
CC(C)C(C(C)(C)COC(=O)C(C)C)O
216.32
16 mPa.s (20°C)
98+%
2,2,4-Trimethyl-1,3-pentanediolmono(2-methylpropanoate)
98.0
0.9500g/mL
114°C
98% min. (sum two isomers) (GC)
C12H24O3
MFCD00148967
0.95
DAFHKNAQFPVRKR-UHFFFAOYSA-N
(3-hydroxy-2,2,4-trimethylpentyl) 2-methylpropanoate
6490
216.32
Liquid
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 246-771-9

RTECSNumber : UF6000000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only