Learn More
2,2'-Azobis(2-methylpropionamidine) dihydrochloride, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 401560250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Radical initiatorsSpecifications
2,2′-Azobis(2-methylpropionamidine) dihydrochloride | |
97.5 | |
White | |
1% max. | |
97.5 to 102.5% (on dry substance) (Argentometry) | |
C8H18N6·2HCl | |
25 g | |
aaph, 2,2'-diazene-1,2-diyl bis 2-methylpropanimidamide dihydrochloride, 2,2'-azobis 2-methylpropionamidine dihydrochloride, ms 1 catalyst, 2,2'-azobis 2-amidinopropane dihydrochloride, unii-7381jdr72f, ccris 7279, ms 1, propanimidamide, 2,2'-azobis 2-methyl-, dihydrochloride, abap | |
LXEKPEMOWBOYRF-UHFFFAOYSA-N | |
2-[(1-amino-1-imino-2-methylpropan-2-yl)diazenyl]-2-methylpropanimidamide;dihydrochloride | |
76344 | |
98% |
2997-92-4 | |
100.0 | |
0.4200g/mL | |
Authentic | |
Plastic bottle | |
MFCD00142725 | |
04,IV,3379 | |
Solubility in water: soluble. | |
CC(C)(C(=N)N)N=NC(C)(C)C(=N)N.Cl.Cl | |
271.19 | |
271.19 | |
Granules |
Chemical Identifiers
2997-92-4 | |
271.19 | |
LXEKPEMOWBOYRF-UHFFFAOYSA-N | |
76344 | |
CC(C)(C(=N)N)N=NC(C)(C)C(=N)N.Cl.Cl |
C8H18N6·2HCl | |
MFCD00142725 | |
aaph, 2,2'-diazene-1,2-diyl bis 2-methylpropanimidamide dihydrochloride, 2,2'-azobis 2-methylpropionamidine dihydrochloride, ms 1 catalyst, 2,2'-azobis 2-amidinopropane dihydrochloride, unii-7381jdr72f, ccris 7279, ms 1, propanimidamide, 2,2'-azobis 2-methyl-, dihydrochloride, abap | |
2-[(1-amino-1-imino-2-methylpropan-2-yl)diazenyl]-2-methylpropanimidamide;dihydrochloride |
Safety and Handling
GHS H Statement
Self-heating; may catch fire.
Harmful if swallowed.
May cause an allergic skin reaction.
Toxic to aquatic life with long lasting effects.
GHS P Statement
Keep cool.
Protect from sunlight.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation or rash occurs: Get medical advice/at
GHS Signal Word: Danger
EINECSNumber : 221-070-
RUO – Research Use Only