Learn More
2,2'-Azobis(2-methylpropionamidine) dihydrochloride, 98%, Thermo Scientific Chemicals
$84.24 - $348.52
Chemical Identifiers
CAS | 2997-92-4 |
---|---|
Molecular Formula | C8H18N6·2HCl |
Molecular Weight (g/mol) | 271.19 |
MDL Number | MFCD00142725 |
InChI Key | LXEKPEMOWBOYRF-UHFFFAOYSA-N |
Synonym | aaph, 2,2'-diazene-1,2-diyl bis 2-methylpropanimidamide dihydrochloride, 2,2'-azobis 2-methylpropionamidine dihydrochloride, ms 1 catalyst, 2,2'-azobis 2-amidinopropane dihydrochloride, unii-7381jdr72f, ccris 7279, ms 1, propanimidamide, 2,2'-azobis 2-methyl-, dihydrochloride, abap |
PubChem CID | 76344 |
IUPAC Name | 2-[(1-amino-1-imino-2-methylpropan-2-yl)diazenyl]-2-methylpropanimidamide;dihydrochloride |
SMILES | CC(C)(C(=N)N)N=NC(C)(C)C(=N)N.Cl.Cl |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC401560250
|
Thermo Scientific Chemicals
401560250 |
25 g | Plastic bottle |
Each for $84.24
|
|
||||
AC401560500
|
Thermo Scientific Chemicals
401560500 |
50 g | Plastic Bottle |
Each for $117.90
|
|
||||
AC401562500
|
Thermo Scientific Chemicals
401562500 |
250 g | Plastic bottle |
Each for $348.52
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Radical initiatorsChemical Identifiers
2997-92-4 | |
271.19 | |
LXEKPEMOWBOYRF-UHFFFAOYSA-N | |
76344 | |
CC(C)(C(=N)N)N=NC(C)(C)C(=N)N.Cl.Cl |
C8H18N6·2HCl | |
MFCD00142725 | |
aaph, 2,2'-diazene-1,2-diyl bis 2-methylpropanimidamide dihydrochloride, 2,2'-azobis 2-methylpropionamidine dihydrochloride, ms 1 catalyst, 2,2'-azobis 2-amidinopropane dihydrochloride, unii-7381jdr72f, ccris 7279, ms 1, propanimidamide, 2,2'-azobis 2-methyl-, dihydrochloride, abap | |
2-[(1-amino-1-imino-2-methylpropan-2-yl)diazenyl]-2-methylpropanimidamide;dihydrochloride |
Specifications
2997-92-4 | |
100.0 | |
0.4200g/mL | |
Authentic | |
Plastic bottle | |
MFCD00142725 | |
04,IV,3379 | |
Solubility in water: soluble. | |
CC(C)(C(=N)N)N=NC(C)(C)C(=N)N.Cl.Cl | |
271.19 | |
271.19 | |
Granules |
97.5 | |
White | |
1% max. | |
97.5 to 102.5% (on dry substance) (Argentometry) | |
C8H18N6·2HCl | |
25 g | |
aaph, 2,2'-diazene-1,2-diyl bis 2-methylpropanimidamide dihydrochloride, 2,2'-azobis 2-methylpropionamidine dihydrochloride, ms 1 catalyst, 2,2'-azobis 2-amidinopropane dihydrochloride, unii-7381jdr72f, ccris 7279, ms 1, propanimidamide, 2,2'-azobis 2-methyl-, dihydrochloride, abap | |
LXEKPEMOWBOYRF-UHFFFAOYSA-N | |
2-[(1-amino-1-imino-2-methylpropan-2-yl)diazenyl]-2-methylpropanimidamide;dihydrochloride | |
76344 | |
98% | |
2,2′-Azobis(2-methylpropionamidine) dihydrochloride |
Safety and Handling
GHS H Statement
Self-heating; may catch fire.
Harmful if swallowed.
May cause an allergic skin reaction.
Toxic to aquatic life with long lasting effects.
GHS P Statement
Keep cool.
Protect from sunlight.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation or rash occurs: Get medical advice/at
GHS Signal Word: Danger
EINECSNumber : 221-070-
RUO – Research Use Only