missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,2'-Azobis(2-methylpropionamidine) dihydrochloride, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 401562500

 View more versions of this product

Catalog No. AC401562500


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Radical initiators
Specifications

Specifications

2,2′-Azobis(2-methylpropionamidine) dihydrochloride
97.5
White
1% max.
97.5 to 102.5% (on dry substance) (Argentometry)
C8H18N6·2HCl
250 g
aaph, 2,2'-diazene-1,2-diyl bis 2-methylpropanimidamide dihydrochloride, 2,2'-azobis 2-methylpropionamidine dihydrochloride, ms 1 catalyst, 2,2'-azobis 2-amidinopropane dihydrochloride, unii-7381jdr72f, ccris 7279, ms 1, propanimidamide, 2,2'-azobis 2-methyl-, dihydrochloride, abap
LXEKPEMOWBOYRF-UHFFFAOYSA-N
2-[(1-amino-1-imino-2-methylpropan-2-yl)diazenyl]-2-methylpropanimidamide;dihydrochloride
76344
98%
2997-92-4
100.0
0.4200g/mL
Authentic
Plastic bottle
MFCD00142725
04,IV,3379
Solubility in water: soluble.
CC(C)(C(=N)N)N=NC(C)(C)C(=N)N.Cl.Cl
271.19
271.19
Granules

Chemical Identifiers

2997-92-4
271.19
LXEKPEMOWBOYRF-UHFFFAOYSA-N
76344
CC(C)(C(=N)N)N=NC(C)(C)C(=N)N.Cl.Cl
C8H18N6·2HCl
MFCD00142725
aaph, 2,2'-diazene-1,2-diyl bis 2-methylpropanimidamide dihydrochloride, 2,2'-azobis 2-methylpropionamidine dihydrochloride, ms 1 catalyst, 2,2'-azobis 2-amidinopropane dihydrochloride, unii-7381jdr72f, ccris 7279, ms 1, propanimidamide, 2,2'-azobis 2-methyl-, dihydrochloride, abap
2-[(1-amino-1-imino-2-methylpropan-2-yl)diazenyl]-2-methylpropanimidamide;dihydrochloride
Safety and Handling

Safety and Handling

GHS H Statement
Self-heating; may catch fire.
Harmful if swallowed.
May cause an allergic skin reaction.
Toxic to aquatic life with long lasting effects.

GHS P Statement
Keep cool.
Protect from sunlight.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation or rash occurs: Get medical advice/at

GHS Signal Word: Danger

EINECSNumber : 221-070-

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only