Learn More
2,2'-Bipyridine-4,4'-dicarboxylic acid, 98%
CAS: 6813-38-3 | C12H6N2O4 | 242.19 g/mol
Supplier: Thermo Scientific Chemicals B2259403
Description
2,2'-Bipyridine-4,4'-dicarboxylic acid is used as an organic chemical synthesis intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2,2′-Bipyridine-4,4′-dicarboxylic acid is used as an organic chemical synthesis intermediate.
Solubility
Insoluble in water. Soluble in common organic solvents.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
| 2,2'-Bipyridine-4,4'-dicarboxylic acid | |
| >310°C | |
| MFCD00015430 | |
| 2,2'-bipyridine-4,4'-dicarboxylic acid, 2,2'-bipyridyl-4,4'-dicarboxylic acid, 4,4'-dicarboxy-2,2'-bipyridine, 2,2'-biisonicotinic acid, 2,2'-dipyridyl-4,4'-dicarboxylic acid, 2,2'-bipyridine-4,4'-dicarboxylicacid, 2-4-carboxypyridin-2-yl pyridine-4-carboxylic acid, 4,4-dicarboxy-2,2-bipyridine, 2-4-carboxy-2-pyridyl pyridine-4-carboxylic acid | |
| FXPLCAKVOYHAJA-UHFFFAOYSA-L | |
| 2-(4-carboxypyridin-2-yl)pyridine-4-carboxylic acid | |
| 688094 | |
| 98% |
| 6813-38-3 | |
| C12H6N2O4 | |
| 1 g | |
| Insoluble in water. Soluble in common organic solvents. | |
| [O-]C(=O)C1=CC=NC(=C1)C1=CC(=CC=N1)C([O-])=O | |
| 242.19 | |
| 244.21 |
Chemical Identifiers
| 6813-38-3 | |
| 242.19 | |
| FXPLCAKVOYHAJA-UHFFFAOYSA-L | |
| 688094 | |
| [O-]C(=O)C1=CC=NC(=C1)C1=CC(=CC=N1)C([O-])=O |
| C12H6N2O4 | |
| MFCD00015430 | |
| 2,2'-bipyridine-4,4'-dicarboxylic acid, 2,2'-bipyridyl-4,4'-dicarboxylic acid, 4,4'-dicarboxy-2,2'-bipyridine, 2,2'-biisonicotinic acid, 2,2'-dipyridyl-4,4'-dicarboxylic acid, 2,2'-bipyridine-4,4'-dicarboxylicacid, 2-4-carboxypyridin-2-yl pyridine-4-carboxylic acid, 4,4-dicarboxy-2,2-bipyridine, 2-4-carboxy-2-pyridyl pyridine-4-carboxylic acid | |
| 2-(4-carboxypyridin-2-yl)pyridine-4-carboxylic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only