missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,2'-Bipyridine-4,4'-dicarboxylic acid, 98%, Thermo Scientific Chemicals

$142.81 - $2465.34

Chemical Identifiers

CAS 6813-38-3
Molecular Formula C12H6N2O4
Molecular Weight (g/mol) 242.19
MDL Number MFCD00015430
InChI Key FXPLCAKVOYHAJA-UHFFFAOYSA-L
Synonym 2,2'-bipyridine-4,4'-dicarboxylic acid, 2,2'-bipyridyl-4,4'-dicarboxylic acid, 4,4'-dicarboxy-2,2'-bipyridine, 2,2'-biisonicotinic acid, 2,2'-dipyridyl-4,4'-dicarboxylic acid, 2,2'-bipyridine-4,4'-dicarboxylicacid, 2-4-carboxypyridin-2-yl pyridine-4-carboxylic acid, 4,4-dicarboxy-2,2-bipyridine, 2-4-carboxy-2-pyridyl pyridine-4-carboxylic acid
PubChem CID 688094
IUPAC Name 2-(4-carboxypyridin-2-yl)pyridine-4-carboxylic acid
SMILES [O-]C(=O)C1=CC=NC(=C1)C1=CC(=CC=N1)C([O-])=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2259403
View Documents
Thermo Scientific Chemicals
B2259403
1 g
Each for $142.81
 
AAB2259406
View Documents
Thermo Scientific Chemicals
B2259406
5 g
Each for $522.29
 
AAB2259414
View Documents
Thermo Scientific Chemicals
B2259414
25 g
Each for $2,465.34
 
Description

Description

2,2'-Bipyridine-4,4'-dicarboxylic acid is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2′-Bipyridine-4,4′-dicarboxylic acid is used as an organic chemical synthesis intermediate.

Solubility
Insoluble in water. Soluble in common organic solvents.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

6813-38-3
242.19
FXPLCAKVOYHAJA-UHFFFAOYSA-L
688094
[O-]C(=O)C1=CC=NC(=C1)C1=CC(=CC=N1)C([O-])=O
C12H6N2O4
MFCD00015430
2,2'-bipyridine-4,4'-dicarboxylic acid, 2,2'-bipyridyl-4,4'-dicarboxylic acid, 4,4'-dicarboxy-2,2'-bipyridine, 2,2'-biisonicotinic acid, 2,2'-dipyridyl-4,4'-dicarboxylic acid, 2,2'-bipyridine-4,4'-dicarboxylicacid, 2-4-carboxypyridin-2-yl pyridine-4-carboxylic acid, 4,4-dicarboxy-2,2-bipyridine, 2-4-carboxy-2-pyridyl pyridine-4-carboxylic acid
2-(4-carboxypyridin-2-yl)pyridine-4-carboxylic acid

Specifications

6813-38-3
C12H6N2O4
1 g
Insoluble in water. Soluble in common organic solvents.
[O-]C(=O)C1=CC=NC(=C1)C1=CC(=CC=N1)C([O-])=O
242.19
244.21
2,2'-Bipyridine-4,4'-dicarboxylic acid
>310°C
MFCD00015430
2,2'-bipyridine-4,4'-dicarboxylic acid, 2,2'-bipyridyl-4,4'-dicarboxylic acid, 4,4'-dicarboxy-2,2'-bipyridine, 2,2'-biisonicotinic acid, 2,2'-dipyridyl-4,4'-dicarboxylic acid, 2,2'-bipyridine-4,4'-dicarboxylicacid, 2-4-carboxypyridin-2-yl pyridine-4-carboxylic acid, 4,4-dicarboxy-2,2-bipyridine, 2-4-carboxy-2-pyridyl pyridine-4-carboxylic acid
FXPLCAKVOYHAJA-UHFFFAOYSA-L
2-(4-carboxypyridin-2-yl)pyridine-4-carboxylic acid
688094
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only