missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,2'-Bipyridine-4,4'-dicarboxylic acid, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2259414

 View more versions of this product

Catalog No. AAB2259414


Explore available promotions

Description

Description

2,2'-Bipyridine-4,4'-dicarboxylic acid is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2′-Bipyridine-4,4′-dicarboxylic acid is used as an organic chemical synthesis intermediate.

Solubility
Insoluble in water. Soluble in common organic solvents.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Specifications

2,2'-Bipyridine-4,4'-dicarboxylic acid
>310°C
MFCD00015430
2,2'-bipyridine-4,4'-dicarboxylic acid, 2,2'-bipyridyl-4,4'-dicarboxylic acid, 4,4'-dicarboxy-2,2'-bipyridine, 2,2'-biisonicotinic acid, 2,2'-dipyridyl-4,4'-dicarboxylic acid, 2,2'-bipyridine-4,4'-dicarboxylicacid, 2-4-carboxypyridin-2-yl pyridine-4-carboxylic acid, 4,4-dicarboxy-2,2-bipyridine, 2-4-carboxy-2-pyridyl pyridine-4-carboxylic acid
FXPLCAKVOYHAJA-UHFFFAOYSA-L
2-(4-carboxypyridin-2-yl)pyridine-4-carboxylic acid
688094
98%
6813-38-3
C12H6N2O4
25 g
Insoluble in water. Soluble in common organic solvents.
[O-]C(=O)C1=CC=NC(=C1)C1=CC(=CC=N1)C([O-])=O
242.19
244.21

Chemical Identifiers

6813-38-3
242.19
FXPLCAKVOYHAJA-UHFFFAOYSA-L
688094
[O-]C(=O)C1=CC=NC(=C1)C1=CC(=CC=N1)C([O-])=O
C12H6N2O4
MFCD00015430
2,2'-bipyridine-4,4'-dicarboxylic acid, 2,2'-bipyridyl-4,4'-dicarboxylic acid, 4,4'-dicarboxy-2,2'-bipyridine, 2,2'-biisonicotinic acid, 2,2'-dipyridyl-4,4'-dicarboxylic acid, 2,2'-bipyridine-4,4'-dicarboxylicacid, 2-4-carboxypyridin-2-yl pyridine-4-carboxylic acid, 4,4-dicarboxy-2,2-bipyridine, 2-4-carboxy-2-pyridyl pyridine-4-carboxylic acid
2-(4-carboxypyridin-2-yl)pyridine-4-carboxylic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only