missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,2-Bithiophene-5-carboxylic acid, 97%, Thermo Scientific™

$84.11 - $224.30

Chemical Identifiers

CAS 2060-55-1
Molecular Formula C9H6O2S2
Molecular Weight (g/mol) 210.265
InChI Key PTNWHEHDBNNKEO-UHFFFAOYSA-N
Synonym 2,2'-bithiophene-5-carboxylic acid, 5-thiophen-2-yl thiophene-2-carboxylic acid, 2,2-bithiophene-5-carboxylic acid, 2,2'-bithiophene-5-carboxylicacid, 2,2' bithiophenyl-5-carboxylic acid, 2,2'-bithiophene-5-ca, 5-carboxy-2,2'-bithiophene, 2,2'-bithienyl-5'-carboxylic, 5-2-thienyl-2-thiophenecarboxylic acid
PubChem CID 150965
IUPAC Name 5-thiophen-2-ylthiophene-2-carboxylic acid
SMILES C1=CSC(=C1)C2=CC=C(S2)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
CC14601CB
SDS View Documents
Thermo Scientific Chemicals
CC14601CB
250 mg
Each for $84.11
 
CC14601DA
SDS View Documents
Thermo Scientific Chemicals
CC14601DA
1 g
Each for $224.30
 
Description

Description

Specifications

Chemical Identifiers

2060-55-1
210.265
2,2'-bithiophene-5-carboxylic acid, 5-thiophen-2-yl thiophene-2-carboxylic acid, 2,2-bithiophene-5-carboxylic acid, 2,2'-bithiophene-5-carboxylicacid, 2,2' bithiophenyl-5-carboxylic acid, 2,2'-bithiophene-5-ca, 5-carboxy-2,2'-bithiophene, 2,2'-bithienyl-5'-carboxylic, 5-2-thienyl-2-thiophenecarboxylic acid
5-thiophen-2-ylthiophene-2-carboxylic acid
C9H6O2S2
PTNWHEHDBNNKEO-UHFFFAOYSA-N
150965
C1=CSC(=C1)C2=CC=C(S2)C(=O)O

Specifications

2060-55-1
C9H6O2S2
2,2'-bithiophene-5-carboxylic acid, 5-thiophen-2-yl thiophene-2-carboxylic acid, 2,2-bithiophene-5-carboxylic acid, 2,2'-bithiophene-5-carboxylicacid, 2,2' bithiophenyl-5-carboxylic acid, 2,2'-bithiophene-5-ca, 5-carboxy-2,2'-bithiophene, 2,2'-bithienyl-5'-carboxylic, 5-2-thienyl-2-thiophenecarboxylic acid
C1=CSC(=C1)C2=CC=C(S2)C(=O)O
210.265
210.28
2, 2'-Bithiophene-5-carboxylic acid
97%
250 mg
PTNWHEHDBNNKEO-UHFFFAOYSA-N
5-thiophen-2-ylthiophene-2-carboxylic acid
150965
97%
Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'