Learn More
2,2-Difluoro-4-pentenoic acid, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H3217306
Description
2,2-Difluoro-4-pentenoic acid is involved in the iodolactonization reaction to give the corresponding gamma- lactone, which is converted to prepare prepare 5-hydroxy-3,3-difluoropiperidine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2,2-Difluoro-4-pentenoic acid is involved in the iodolactonization reaction to give the corresponding gamma- lactone, which is converted to prepare prepare 5-hydroxy-3,3-difluoropiperidine.
Solubility
Immiscible with water.
Notes
Incompatible with oxidizing agents and bases.
Specifications
2,2-Difluoro-4-pentenoic acid | |
56°C to 58°C (3 mmHg) | |
MFCD09800642 | |
UN3265 | |
Immiscible with water. | |
C=CCC(C(=O)O)(F)F | |
136.098 | |
136.1 |
55039-89-9 | |
C5H6F2O2 | |
5 g | |
2,2-difluoropent-4-enoicacid, 2,2-difluoro-4-pentenoic acid, 4-pentenoic acid, 2,2-difluoro, 2,2-difluoro-pent-4-enoic acid, 10.14272/lhokyuduayxfgf-uhfffaoysa-n, doi:10.14272/lhokyuduayxfgf-uhfffaoysa-n | |
LHOKYUDUAYXFGF-UHFFFAOYSA-N | |
2,2-difluoropent-4-enoic acid | |
11954656 | |
97% |
Chemical Identifiers
55039-89-9 | |
136.098 | |
LHOKYUDUAYXFGF-UHFFFAOYSA-N | |
11954656 | |
C=CCC(C(=O)O)(F)F |
C5H6F2O2 | |
MFCD09800642 | |
2,2-difluoropent-4-enoicacid, 2,2-difluoro-4-pentenoic acid, 4-pentenoic acid, 2,2-difluoro, 2,2-difluoro-pent-4-enoic acid, 10.14272/lhokyuduayxfgf-uhfffaoysa-n, doi:10.14272/lhokyuduayxfgf-uhfffaoysa-n | |
2,2-difluoropent-4-enoic acid |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302-H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only