missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,2-Dimethoxy-2-phenylacetophenone, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 187840500

 View more versions of this product

Catalog No. AC187840500


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2, 2-Dimethoxy-2-phenylacetophenone
24650-42-8
100.0
White to Yellow
Authentic
Glass bottle
C6H5COC(OCH3)2C6H5
50 g
2,2-dimethoxy-2-phenylacetophenone, benzil dimethyl ketal, kayacure bdmk, irgacure 651, ethanone, 2,2-dimethoxy-1,2-diphenyl, lucirin bdk, photomer 51, benzil dimethylketal, esacure kb 1, irgacure 621
COC(OC)(C(=O)C1=CC=CC=C1)C1=CC=CC=C1
256.30
256.29
Crystalline Powder
99%
98.5
64°C to 68°C
0.2% max. (75°C, 2 hrs)
98.5% min. (GC)
C16H16O3
MFCD00008475
14, 3392
KWVGIHKZDCUPEU-UHFFFAOYSA-N
2,2-dimethoxy-1,2-diphenylethanone
90571
99%

Chemical Identifiers

24650-42-8
256.30
KWVGIHKZDCUPEU-UHFFFAOYSA-N
90571
COC(OC)(C(=O)C1=CC=CC=C1)C1=CC=CC=C1
C16H16O3
MFCD00008475
2,2-dimethoxy-2-phenylacetophenone, benzil dimethyl ketal, kayacure bdmk, irgacure 651, ethanone, 2,2-dimethoxy-1,2-diphenyl, lucirin bdk, photomer 51, benzil dimethylketal, esacure kb 1, irgacure 621
2,2-dimethoxy-1,2-diphenylethanone
Safety and Handling

Safety and Handling

LIGHT SENSITIVE

EINECSNumber : 246-386-6

RTECSNumber : KM5775658

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only