missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,2-Dimethyl-1,3-dioxolane, 98+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L0048206

 View more versions of this product

Catalog No. AAL0048206


Explore available promotions

Description

Description

2,2-Dimethyl-1,3-dioxolane is used as organic intermediates, fine chemicals, pharmaceutical research and development.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2-Dimethyl-1,3-dioxolane is used as organic intermediates, fine chemicals, pharmaceutical research and development.

Solubility
Insoluble in water.

Notes
Protect from heat. Store away from oxidizing agents.
Specifications

Specifications

2,2-Dimethyl-1,3-dioxolane
0.946
6°C (42°F)
1.398
5 g
102775
Insoluble in water.
CC1(C)OCCO1
102.13
102.13
2916-31-6
91°C to 93°C
C5H10O2
MFCD00014106
UN1993
1,3-dioxolane, 2,2-dimethyl, 2,2-dimethyl-1,3-dioxolan, 1,3-dioxolane, dimethyl, acetone ethylene ketal, acetone ethylene acetal, acetone, cyclic ethylene acetal, 1,3-dioxolane,2,2-dimethyl, sijbdwpvnayvgy-uhfffaoysa, 2,2-dimethyl-1,3-dioxolane
SIJBDWPVNAYVGY-UHFFFAOYSA-N
2,2-dimethyl-1,3-dioxolane
76209
≥98%

Chemical Identifiers

2916-31-6
102.13
SIJBDWPVNAYVGY-UHFFFAOYSA-N
2,2-dimethyl-1,3-dioxolane
C5H10O2
MFCD00014106
76209
CC1(C)OCCO1
Safety and Handling

Safety and Handling

GHS H Statement
H225
Highly flammable liquid and vapor.

P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c

H225

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

EINECSNumber : 220-842-4

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only