Learn More
2,2-Dimethyl-1,3-dioxolane, 98+%, Thermo Scientific Chemicals
$56.77 - $176.38
Chemical Identifiers
CAS | 2916-31-6 |
---|---|
Molecular Formula | C5H10O2 |
Molecular Weight (g/mol) | 102.13 |
MDL Number | MFCD00014106 |
InChI Key | SIJBDWPVNAYVGY-UHFFFAOYSA-N |
PubChem CID | 76209 |
IUPAC Name | 2,2-dimethyl-1,3-dioxolane |
SMILES | CC1(C)OCCO1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL0048206
|
Thermo Scientific Chemicals
L0048206 |
5 g |
Each for $56.77
|
|
|||||
AAL0048214
|
Thermo Scientific Chemicals
L0048214 |
25 g |
Each for $176.38
|
|
|||||
Description
2,2-Dimethyl-1,3-dioxolane is used as organic intermediates, fine chemicals, pharmaceutical research and development.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2,2-Dimethyl-1,3-dioxolane is used as organic intermediates, fine chemicals, pharmaceutical research and development.
Solubility
Insoluble in water.
Notes
Protect from heat. Store away from oxidizing agents.
Chemical Identifiers
2916-31-6 | |
102.13 | |
SIJBDWPVNAYVGY-UHFFFAOYSA-N | |
2,2-dimethyl-1,3-dioxolane |
C5H10O2 | |
MFCD00014106 | |
76209 | |
CC1(C)OCCO1 |
Specifications
2916-31-6 | |
91°C to 93°C | |
C5H10O2 | |
MFCD00014106 | |
UN1993 | |
1,3-dioxolane, 2,2-dimethyl, 2,2-dimethyl-1,3-dioxolan, 1,3-dioxolane, dimethyl, acetone ethylene ketal, acetone ethylene acetal, acetone, cyclic ethylene acetal, 1,3-dioxolane,2,2-dimethyl, sijbdwpvnayvgy-uhfffaoysa, 2,2-dimethyl-1,3-dioxolane | |
SIJBDWPVNAYVGY-UHFFFAOYSA-N | |
2,2-dimethyl-1,3-dioxolane | |
76209 | |
≥98% |
0.946 | |
6°C (42°F) | |
1.398 | |
5 g | |
102775 | |
Insoluble in water. | |
CC1(C)OCCO1 | |
102.13 | |
102.13 | |
2,2-Dimethyl-1,3-dioxolane |
Safety and Handling
GHS H Statement
H225
Highly flammable liquid and vapor.
P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
H225
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber : 220-842-4
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only