missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-[2-(Dimethylamino)ethoxy]ethanol 98.0+%, TCI America™
Supplier: TCI America D175625ML
Specifications
2-[2-(Dimethylamino)ethoxy]ethanol | |
Yellow | |
C6H15NO2 | |
25 mL | |
YSAANLSYLSUVHB-UHFFFAOYSA-N | |
2-[2-(dimethylamino)ethoxy]ethan-1-ol | |
74348 | |
≥98.0% (GC) |
1704-62-7 | |
95°C | |
MFCD00059602 | |
Ethylene Glycol Mono[2-(dimethylamino)ethyl] Ether | |
CN(C)CCOCCO | |
133.19 | |
133.19 | |
Liquid |
Chemical Identifiers
1704-62-7 | |
133.19 | |
YSAANLSYLSUVHB-UHFFFAOYSA-N | |
74348 | |
CN(C)CCOCCO |
C6H15NO2 | |
MFCD00059602 | |
Ethylene Glycol Mono[2-(dimethylamino)ethyl] Ether | |
2-[2-(dimethylamino)ethoxy]ethan-1-ol |
Safety and Handling
RTECSNumber : KK6825000
TSCA : Yes