missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-[2-(Dimethylamino)ethoxy]ethanol 98.0+%, TCI America™
$31.68 - $182.68
Chemical Identifiers
| CAS | 1704-62-7 |
|---|---|
| Molecular Formula | C6H15NO2 |
| Molecular Weight (g/mol) | 133.19 |
| MDL Number | MFCD00059602 |
| InChI Key | YSAANLSYLSUVHB-UHFFFAOYSA-N |
| Synonym | Ethylene Glycol Mono[2-(dimethylamino)ethyl] Ether |
| PubChem CID | 74348 |
| IUPAC Name | 2-[2-(dimethylamino)ethoxy]ethan-1-ol |
| SMILES | CN(C)CCOCCO |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
D175625ML
|
TCI America
D175625ML |
25 mL |
Each for $31.68
|
|
|||||
|
D1756500ML
|
TCI America
D1756500ML |
500 mL |
Each for $182.68
|
|
|||||
Chemical Identifiers
| 1704-62-7 | |
| 133.19 | |
| YSAANLSYLSUVHB-UHFFFAOYSA-N | |
| 74348 | |
| CN(C)CCOCCO |
| C6H15NO2 | |
| MFCD00059602 | |
| Ethylene Glycol Mono[2-(dimethylamino)ethyl] Ether | |
| 2-[2-(dimethylamino)ethoxy]ethan-1-ol |
Specifications
| 1704-62-7 | |
| 95°C | |
| MFCD00059602 | |
| Ethylene Glycol Mono[2-(dimethylamino)ethyl] Ether | |
| CN(C)CCOCCO | |
| 133.19 | |
| 133.19 | |
| Liquid |
| Yellow | |
| C6H15NO2 | |
| 25 mL | |
| YSAANLSYLSUVHB-UHFFFAOYSA-N | |
| 2-[2-(dimethylamino)ethoxy]ethan-1-ol | |
| 74348 | |
| ≥98.0% (GC) | |
| 2-[2-(Dimethylamino)ethoxy]ethanol |
Safety and Handling
RTECSNumber : KK6825000
TSCA : Yes