missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(2-Ethoxyethoxy)ethanol, 99-100%, Spectrum™ Chemical
$259.68 - $15879.50
Chemical Identifiers
| CAS | 111-90-0 |
|---|---|
| Molecular Formula | C6H14O3 |
| Molecular Weight (g/mol) | 134.18 |
| InChI Key | XXJWXESWEXIICW-UHFFFAOYSA-N |
| IUPAC Name | 2-(2-ethoxyethoxy)ethan-1-ol |
| SMILES | CCOCCOCCO |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18608147
|
Spectrum Chemical Mfg Cor
E10221LT |
1 L | Amber Glass Bottle |
Each for $259.68
|
|
||||
|
18608148
|
Spectrum Chemical Mfg Cor
E10224LTGL |
4 L | Amber Glass Bottle |
Each for $774.47
|
|
||||
|
18608144
|
Spectrum Chemical Mfg Cor
E102220LTMT |
20 L | Steel Can |
Each for $2,619.20
|
|
||||
|
18608145
|
Spectrum Chemical Mfg Cor
E1022200LTBL |
200 L | Drum |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.