missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,3-Butanediol, 98%, mixture of racemic and meso forms, techn., Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 107642500

 View more versions of this product

Catalog No. AC107642500


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2, 3-Butanediol
19.0°C
1.0100g/mL
85°C
97.5% min. (GC)
C4H10O2
CH3CH(OH)CH(OH)CH3
01,479
15,1571
Solubility in water: soluble in water. Other solubilities: soluble in alcohol, ether and diisopropyl ether
CC(C(C)O)O
90.12
CHEBI:62064
98%
513-85-9
Colorless to Yellow
180.0°C
Authentic
Glass bottle
1.4330 to 1.4370
250 mL
1.01
2,3-butanediol, 2,3-butylene glycol, 2,3-dihydroxybutane, dimethylethylene glycol, dimethylene glycol, pseudobutylene glycol, sym-dimethylethylene glycol, ccris 5501, dl-2,3-butanediol, 2,3-butanediol, r*,r*-.+/-.
OWBTYPJTUOEWEK-UHFFFAOYSA-N
butane-2,3-diol
262
90.12
Viscous Liquid

Chemical Identifiers

513-85-9
90.12
2,3-butanediol, 2,3-butylene glycol, 2,3-dihydroxybutane, dimethylethylene glycol, dimethylene glycol, pseudobutylene glycol, sym-dimethylethylene glycol, ccris 5501, dl-2,3-butanediol, 2,3-butanediol, r*,r*-.+/-.
CHEBI:62064
CC(C(C)O)O
C4H10O2
OWBTYPJTUOEWEK-UHFFFAOYSA-N
262
butane-2,3-diol
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 208-173-6

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only