missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2,3-Butanediol, 98%, mixture of racemic and meso forms, techn., Thermo Scientific Chemicals
$516.42 - $516.42
Chemical Identifiers
CAS | 513-85-9 |
---|---|
Molecular Formula | C4H10O2 |
Molecular Weight (g/mol) | 90.12 |
InChI Key | OWBTYPJTUOEWEK-UHFFFAOYSA-N |
Synonym | 2,3-butanediol, 2,3-butylene glycol, 2,3-dihydroxybutane, dimethylethylene glycol, dimethylene glycol, pseudobutylene glycol, sym-dimethylethylene glycol, ccris 5501, dl-2,3-butanediol, 2,3-butanediol, r*,r*-.+/-. |
PubChem CID | 262 |
ChEBI | CHEBI:62064 |
IUPAC Name | butane-2,3-diol |
SMILES | CC(C(C)O)O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC107642500
|
Thermo Scientific Chemicals
107642500 |
250 mL | Glass bottle |
Each for $516.42
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
513-85-9 | |
90.12 | |
2,3-butanediol, 2,3-butylene glycol, 2,3-dihydroxybutane, dimethylethylene glycol, dimethylene glycol, pseudobutylene glycol, sym-dimethylethylene glycol, ccris 5501, dl-2,3-butanediol, 2,3-butanediol, r*,r*-.+/-. | |
CHEBI:62064 | |
CC(C(C)O)O |
C4H10O2 | |
OWBTYPJTUOEWEK-UHFFFAOYSA-N | |
262 | |
butane-2,3-diol |
Specifications
513-85-9 | |
Colorless to Yellow | |
180.0°C | |
Authentic | |
Glass bottle | |
1.4330 to 1.4370 | |
250 mL | |
1.01 | |
2,3-butanediol, 2,3-butylene glycol, 2,3-dihydroxybutane, dimethylethylene glycol, dimethylene glycol, pseudobutylene glycol, sym-dimethylethylene glycol, ccris 5501, dl-2,3-butanediol, 2,3-butanediol, r*,r*-.+/-. | |
OWBTYPJTUOEWEK-UHFFFAOYSA-N | |
butane-2,3-diol | |
262 | |
90.12 | |
Viscous Liquid |
19.0°C | |
1.0100g/mL | |
85°C | |
97.5% min. (GC) | |
C4H10O2 | |
CH3CH(OH)CH(OH)CH3 | |
01,479 | |
15,1571 | |
Solubility in water: soluble in water. Other solubilities: soluble in alcohol, ether and diisopropyl ether | |
CC(C(C)O)O | |
90.12 | |
CHEBI:62064 | |
98% | |
2, 3-Butanediol |
Safety and Handling
EINECSNumber : 208-173-6
RUO – Research Use Only