missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,3'-Dibromoacetophenone, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1521006

 View more versions of this product

Catalog No. AAA1521006


Explore available promotions

Description

Description

It is employed as an intermediate for pharmaceutical and organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as an intermediate for pharmaceutical and organic synthesis.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
Specifications

Specifications

2,3'-Dibromoacetophenone
49°C to 52°C
C8H6Br2O
5 g
775786
Insoluble in water.
C1=CC(=CC(=C1)Br)C(=O)CBr
277.943
277.95
18523-22-3
Pungent
MFCD00082698
UN3261
3-bromophenacyl bromide, 2,3'-dibromoacetophenone, 2-bromo-1-3-bromophenyl ethanone, ethanone, 2-bromo-1-3-bromophenyl, 2-bromo-1-3-bromophenyl ethan-1-one, 3-bromophenacylbromide, 2-bromo-1-3-bromo-phenyl-ethanone, 2-bromo-3'-bromoacetophenone, 2-bromo-1-3-bromophenyl-1-ethanone
MZBXSQBQPJWECM-UHFFFAOYSA-N
2-bromo-1-(3-bromophenyl)ethanone
519585
97%

Chemical Identifiers

18523-22-3
277.943
MZBXSQBQPJWECM-UHFFFAOYSA-N
519585
C1=CC(=CC(=C1)Br)C(=O)CBr
C8H6Br2O
MFCD00082698
3-bromophenacyl bromide, 2,3'-dibromoacetophenone, 2-bromo-1-3-bromophenyl ethanone, ethanone, 2-bromo-1-3-bromophenyl, 2-bromo-1-3-bromophenyl ethan-1-one, 3-bromophenacylbromide, 2-bromo-1-3-bromo-phenyl-ethanone, 2-bromo-3'-bromoacetophenone, 2-bromo-1-3-bromophenyl-1-ethanone
2-bromo-1-(3-bromophenyl)ethanone
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314

DOTInformation : Hazard Class: 8; Packaging Group: III

TSCA : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only