missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,3'-Dibromoacetophenone, 97%, Thermo Scientific Chemicals

$148.44 - $488.81

Chemical Identifiers

CAS 18523-22-3
Molecular Formula C8H6Br2O
Molecular Weight (g/mol) 277.943
MDL Number MFCD00082698
InChI Key MZBXSQBQPJWECM-UHFFFAOYSA-N
Synonym 3-bromophenacyl bromide, 2,3'-dibromoacetophenone, 2-bromo-1-3-bromophenyl ethanone, ethanone, 2-bromo-1-3-bromophenyl, 2-bromo-1-3-bromophenyl ethan-1-one, 3-bromophenacylbromide, 2-bromo-1-3-bromo-phenyl-ethanone, 2-bromo-3'-bromoacetophenone, 2-bromo-1-3-bromophenyl-1-ethanone
PubChem CID 519585
IUPAC Name 2-bromo-1-(3-bromophenyl)ethanone
SMILES C1=CC(=CC(=C1)Br)C(=O)CBr
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1521006
View Documents
Thermo Scientific Chemicals
A1521006
5 g
Each for $148.44
 
AAA1521014
View Documents
Thermo Scientific Chemicals
A1521014
25 g
Each for $488.81
 
Description

Description

It is employed as an intermediate for pharmaceutical and organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as an intermediate for pharmaceutical and organic synthesis.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
Specifications

Chemical Identifiers

18523-22-3
277.943
MZBXSQBQPJWECM-UHFFFAOYSA-N
519585
C1=CC(=CC(=C1)Br)C(=O)CBr
C8H6Br2O
MFCD00082698
3-bromophenacyl bromide, 2,3'-dibromoacetophenone, 2-bromo-1-3-bromophenyl ethanone, ethanone, 2-bromo-1-3-bromophenyl, 2-bromo-1-3-bromophenyl ethan-1-one, 3-bromophenacylbromide, 2-bromo-1-3-bromo-phenyl-ethanone, 2-bromo-3'-bromoacetophenone, 2-bromo-1-3-bromophenyl-1-ethanone
2-bromo-1-(3-bromophenyl)ethanone

Specifications

18523-22-3
Pungent
MFCD00082698
UN3261
3-bromophenacyl bromide, 2,3'-dibromoacetophenone, 2-bromo-1-3-bromophenyl ethanone, ethanone, 2-bromo-1-3-bromophenyl, 2-bromo-1-3-bromophenyl ethan-1-one, 3-bromophenacylbromide, 2-bromo-1-3-bromo-phenyl-ethanone, 2-bromo-3'-bromoacetophenone, 2-bromo-1-3-bromophenyl-1-ethanone
MZBXSQBQPJWECM-UHFFFAOYSA-N
2-bromo-1-(3-bromophenyl)ethanone
519585
97%
49°C to 52°C
C8H6Br2O
5 g
775786
Insoluble in water.
C1=CC(=CC(=C1)Br)C(=O)CBr
277.943
277.95
2,3'-Dibromoacetophenone
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314

DOTInformation : Hazard Class: 8; Packaging Group: III

TSCA : No

Recommended Storage : Keep cold

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only