missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2,3-Dimethyl-2,3-diphenylbutane, 95%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 349981000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
2, 3-Dimethyl-2, 3-diphenylbutane | |
1889-67-4 | |
100.0 | |
94% min | |
C18H22 | |
MFCD00053713 | |
05,396 | |
Solubility in water: insoluble in water. Other solubilities: soluble in toluene | |
CC(C)(C1=CC=CC=C1)C(C)(C)C2=CC=CC=C2 | |
238.37 | |
238.37 |
95% | |
94.0 | |
Authentic | |
Glass bottle | |
C6H5C(CH3)2C(CH3)2C6H5 | |
100g | |
2,3-dimethyl-2,3-diphenylbutane, dicumene, 2,3-dimethylbutane-2,3-diyl dibenzene, dicumyl, butane, 2,3-dimethyl-2,3-diphenyl, benzene, 1,1'-1,1,2,2-tetramethyl-1,2-ethanediyl bis, .alpha.,.alpha.'-dicumyl, 2,3-dimethyl-3-phenylbutan-2-yl benzene, bibenzyl, .alpha.,.alpha.,.alpha.',.alpha.'-tetramethyl, 1,1,2-trimethyl-2-phenylpropyl benzene | |
HGTUJZTUQFXBIH-UHFFFAOYSA-N | |
(2,3-dimethyl-3-phenylbutan-2-yl)benzene | |
74681 | |
95% |
Chemical Identifiers
1889-67-4 | |
238.37 | |
HGTUJZTUQFXBIH-UHFFFAOYSA-N | |
74681 | |
CC(C)(C1=CC=CC=C1)C(C)(C)C2=CC=CC=C2 |
C18H22 | |
MFCD00053713 | |
2,3-dimethyl-2,3-diphenylbutane, dicumene, 2,3-dimethylbutane-2,3-diyl dibenzene, dicumyl, butane, 2,3-dimethyl-2,3-diphenyl, benzene, 1,1'-1,1,2,2-tetramethyl-1,2-ethanediyl bis, .alpha.,.alpha.'-dicumyl, 2,3-dimethyl-3-phenylbutan-2-yl benzene, bibenzyl, .alpha.,.alpha.,.alpha.',.alpha.'-tetramethyl, 1,1,2-trimethyl-2-phenylpropyl benzene | |
(2,3-dimethyl-3-phenylbutan-2-yl)benzene |
Safety and Handling
EINECSNumber : 217-568-2
TSCA : TSCA
RUO â Research Use Only