missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2,3-Dimethyl-2,3-diphenylbutane, 95%, Thermo Scientific™
Chemical Identifiers
CAS | 1889-67-4 |
---|---|
Molecular Formula | C18H22 |
Molecular Weight (g/mol) | 238.37 |
MDL Number | MFCD00053713 |
InChI Key | HGTUJZTUQFXBIH-UHFFFAOYSA-N |
Synonym | 2,3-dimethyl-2,3-diphenylbutane, dicumene, 2,3-dimethylbutane-2,3-diyl dibenzene, dicumyl, butane, 2,3-dimethyl-2,3-diphenyl, benzene, 1,1'-1,1,2,2-tetramethyl-1,2-ethanediyl bis, .alpha.,.alpha.'-dicumyl, 2,3-dimethyl-3-phenylbutan-2-yl benzene, bibenzyl, .alpha.,.alpha.,.alpha.',.alpha.'-tetramethyl, 1,1,2-trimethyl-2-phenylpropyl benzene |
PubChem CID | 74681 |
IUPAC Name | (2,3-dimethyl-3-phenylbutan-2-yl)benzene |
SMILES | CC(C)(C1=CC=CC=C1)C(C)(C)C2=CC=CC=C2 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC349980250
|
Thermo Scientific Chemicals
349980250 |
25g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC349981000
|
Thermo Scientific Chemicals
349981000 |
100g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
1889-67-4 | |
238.37 | |
HGTUJZTUQFXBIH-UHFFFAOYSA-N | |
74681 | |
CC(C)(C1=CC=CC=C1)C(C)(C)C2=CC=CC=C2 |
C18H22 | |
MFCD00053713 | |
2,3-dimethyl-2,3-diphenylbutane, dicumene, 2,3-dimethylbutane-2,3-diyl dibenzene, dicumyl, butane, 2,3-dimethyl-2,3-diphenyl, benzene, 1,1'-1,1,2,2-tetramethyl-1,2-ethanediyl bis, .alpha.,.alpha.'-dicumyl, 2,3-dimethyl-3-phenylbutan-2-yl benzene, bibenzyl, .alpha.,.alpha.,.alpha.',.alpha.'-tetramethyl, 1,1,2-trimethyl-2-phenylpropyl benzene | |
(2,3-dimethyl-3-phenylbutan-2-yl)benzene |
Specifications
1889-67-4 | |
100.0 | |
94% min | |
C18H22 | |
MFCD00053713 | |
05,396 | |
Solubility in water: insoluble in water. Other solubilities: soluble in toluene | |
CC(C)(C1=CC=CC=C1)C(C)(C)C2=CC=CC=C2 | |
238.37 | |
238.37 | |
2, 3-Dimethyl-2, 3-diphenylbutane, 95% |
94.0 | |
Authentic | |
Glass bottle | |
C6H5C(CH3)2C(CH3)2C6H5 | |
25g | |
2,3-dimethyl-2,3-diphenylbutane, dicumene, 2,3-dimethylbutane-2,3-diyl dibenzene, dicumyl, butane, 2,3-dimethyl-2,3-diphenyl, benzene, 1,1'-1,1,2,2-tetramethyl-1,2-ethanediyl bis, .alpha.,.alpha.'-dicumyl, 2,3-dimethyl-3-phenylbutan-2-yl benzene, bibenzyl, .alpha.,.alpha.,.alpha.',.alpha.'-tetramethyl, 1,1,2-trimethyl-2-phenylpropyl benzene | |
HGTUJZTUQFXBIH-UHFFFAOYSA-N | |
(2,3-dimethyl-3-phenylbutan-2-yl)benzene | |
74681 | |
95% |
Safety and Handling
EINECSNumber : 217-568-2
TSCA : TSCA
RUO â Research Use Only