Learn More
2-(3-Methoxyphenyl)ethylamine, 97+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6449406
Description
Activated phenethylamine used in the perfluorooctanesulfonic acid catalyzed Pictet-Spengler reaction. Also used in a synthesis of 1,3-oxazepines via palladium-catalyzed intramolecular coupling.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsActivated phenethylamine used in the perfluorooctanesulfonic acid catalyzed Pictet-Spengler reaction. Also used in a synthesis of 1,3-oxazepines via palladium-catalyzed intramolecular coupling.
Solubility
Insoluble in water.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications
2-(3-Methoxyphenyl)ethylamine | |
Yellow | |
118°C to 118°C (6 mmHg) | |
C9H13NO | |
MFCD00008187 | |
UN2735 | |
3-methoxyphenethylamine, 3-methoxyphenylethylamine, 2-3-methoxyphenyl ethanamine, 2-3-methoxyphenyl ethylamine, m-methoxyphenethylamine, m-methoxyphenylethylamine, 2-3-methoxyphenyl ethan-1-amine, benzeneethanamine, 3-methoxy, phenethylamine, m-methoxy, 2-3-methoxy-phenyl-ethylamine | |
WJBMRZAHTUFBGE-UHFFFAOYSA-N | |
2-(3-methoxyphenyl)ethanamine | |
74866 | |
≥97% |
2039-67-0 | |
1.038 g/mL | |
>110°C (230°F) | |
1.538 | |
5 g | |
775202 | |
Insoluble in water. | |
COC1=CC=CC(=C1)CCN | |
151.209 | |
151.21 | |
Liquid |
Chemical Identifiers
2039-67-0 | |
151.209 | |
WJBMRZAHTUFBGE-UHFFFAOYSA-N | |
74866 | |
COC1=CC=CC(=C1)CCN |
C9H13NO | |
MFCD00008187 | |
3-methoxyphenethylamine, 3-methoxyphenylethylamine, 2-3-methoxyphenyl ethanamine, 2-3-methoxyphenyl ethylamine, m-methoxyphenethylamine, m-methoxyphenylethylamine, 2-3-methoxyphenyl ethan-1-amine, benzeneethanamine, 3-methoxy, phenethylamine, m-methoxy, 2-3-methoxy-phenyl-ethylamine | |
2-(3-methoxyphenyl)ethanamine |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314
DOTInformation : Hazard Class: 8; Packaging Group: II
EINECSNumber : 218-017-9
RTECSNumber : SH7835000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only