missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-(3-Methoxyphenyl)ethylamine, 97+%, Thermo Scientific Chemicals

$73.78 - $726.82

Chemical Identifiers

CAS 2039-67-0
Molecular Formula C9H13NO
Molecular Weight (g/mol) 151.209
MDL Number MFCD00008187
InChI Key WJBMRZAHTUFBGE-UHFFFAOYSA-N
Synonym 3-methoxyphenethylamine, 3-methoxyphenylethylamine, 2-3-methoxyphenyl ethanamine, 2-3-methoxyphenyl ethylamine, m-methoxyphenethylamine, m-methoxyphenylethylamine, 2-3-methoxyphenyl ethan-1-amine, benzeneethanamine, 3-methoxy, phenethylamine, m-methoxy, 2-3-methoxy-phenyl-ethylamine
PubChem CID 74866
IUPAC Name 2-(3-methoxyphenyl)ethanamine
SMILES COC1=CC=CC(=C1)CCN
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6449406
View Documents
Thermo Scientific Chemicals
H6449406
5 g
Each for $73.78
 
AAH6449414
View Documents
Thermo Scientific Chemicals
H6449414
25 g
Each for $197.23
 
AAH6449422
View Documents
Thermo Scientific Chemicals
H6449422
100 g
Each for $726.82
 
Description

Description

Activated phenethylamine used in the perfluorooctanesulfonic acid catalyzed Pictet-Spengler reaction. Also used in a synthesis of 1,3-oxazepines via palladium-catalyzed intramolecular coupling.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Activated phenethylamine used in the perfluorooctanesulfonic acid catalyzed Pictet-Spengler reaction. Also used in a synthesis of 1,3-oxazepines via palladium-catalyzed intramolecular coupling.

Solubility
Insoluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications

Chemical Identifiers

2039-67-0
151.209
WJBMRZAHTUFBGE-UHFFFAOYSA-N
74866
COC1=CC=CC(=C1)CCN
C9H13NO
MFCD00008187
3-methoxyphenethylamine, 3-methoxyphenylethylamine, 2-3-methoxyphenyl ethanamine, 2-3-methoxyphenyl ethylamine, m-methoxyphenethylamine, m-methoxyphenylethylamine, 2-3-methoxyphenyl ethan-1-amine, benzeneethanamine, 3-methoxy, phenethylamine, m-methoxy, 2-3-methoxy-phenyl-ethylamine
2-(3-methoxyphenyl)ethanamine

Specifications

2039-67-0
1.038 g/mL
>110°C (230°F)
1.538
5 g
775202
Insoluble in water.
COC1=CC=CC(=C1)CCN
151.209
151.21
Liquid
Yellow
118°C to 118°C (6 mmHg)
C9H13NO
MFCD00008187
UN2735
3-methoxyphenethylamine, 3-methoxyphenylethylamine, 2-3-methoxyphenyl ethanamine, 2-3-methoxyphenyl ethylamine, m-methoxyphenethylamine, m-methoxyphenylethylamine, 2-3-methoxyphenyl ethan-1-amine, benzeneethanamine, 3-methoxy, phenethylamine, m-methoxy, 2-3-methoxy-phenyl-ethylamine
WJBMRZAHTUFBGE-UHFFFAOYSA-N
2-(3-methoxyphenyl)ethanamine
74866
≥97%
2-(3-Methoxyphenyl)ethylamine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314

DOTInformation : Hazard Class: 8; Packaging Group: II

EINECSNumber : 218-017-9

RTECSNumber : SH7835000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only