missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-(3-Methoxyphenyl)ethylamine, 97+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H6449422

 View more versions of this product

Catalog No. AAH6449422


Explore available promotions

Description

Description

Activated phenethylamine used in the perfluorooctanesulfonic acid catalyzed Pictet-Spengler reaction. Also used in a synthesis of 1,3-oxazepines via palladium-catalyzed intramolecular coupling.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Activated phenethylamine used in the perfluorooctanesulfonic acid catalyzed Pictet-Spengler reaction. Also used in a synthesis of 1,3-oxazepines via palladium-catalyzed intramolecular coupling.

Solubility
Insoluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications

Specifications

2-(3-Methoxyphenyl)ethylamine
Yellow
118°C to 118°C (6 mmHg)
C9H13NO
MFCD00008187
UN2735
3-methoxyphenethylamine, 3-methoxyphenylethylamine, 2-3-methoxyphenyl ethanamine, 2-3-methoxyphenyl ethylamine, m-methoxyphenethylamine, m-methoxyphenylethylamine, 2-3-methoxyphenyl ethan-1-amine, benzeneethanamine, 3-methoxy, phenethylamine, m-methoxy, 2-3-methoxy-phenyl-ethylamine
WJBMRZAHTUFBGE-UHFFFAOYSA-N
2-(3-methoxyphenyl)ethanamine
74866
≥97%
2039-67-0
1.038 g/mL
>110°C (230°F)
1.538
100 g
775202
Insoluble in water.
COC1=CC=CC(=C1)CCN
151.209
151.21
Liquid

Chemical Identifiers

2039-67-0
151.209
WJBMRZAHTUFBGE-UHFFFAOYSA-N
74866
COC1=CC=CC(=C1)CCN
C9H13NO
MFCD00008187
3-methoxyphenethylamine, 3-methoxyphenylethylamine, 2-3-methoxyphenyl ethanamine, 2-3-methoxyphenyl ethylamine, m-methoxyphenethylamine, m-methoxyphenylethylamine, 2-3-methoxyphenyl ethan-1-amine, benzeneethanamine, 3-methoxy, phenethylamine, m-methoxy, 2-3-methoxy-phenyl-ethylamine
2-(3-methoxyphenyl)ethanamine
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314

DOTInformation : Hazard Class: 8; Packaging Group: II

EINECSNumber : 218-017-9

RTECSNumber : SH7835000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only