missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-[3-(Trifluoromethyl)phenyl]ethylamine, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H6408406

 View more versions of this product

Catalog No. AAH6408406


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-[3-(Trifluoromethyl)phenyl]ethylamine
Colorless to Yellow
113°C (22 mmHg)
1.4645
5 g
4398117
2-3-trifluoromethyl phenyl ethanamine, 3-trifluoromethyl phenethylamine, 2-3-trifluoromethylphenyl ethylamine, 2-3-trifluoromethyl phenyl ethylamine, phenethylamine, m-trifluoromethyl, m-trifluoromethylphenethylamine, m-trifluoromethyl phenethylamine, benzeneethanamine, 3-trifluoromethyl, 2-3-trifluoromethylphenyl-ethylamine
C1=CC(=CC(=C1)C(F)(F)F)CCN
189.181
189.18
Liquid
52516-30-0
1.18 g/mL
C9H10F3N
MFCD00040756
UN2735
Light sensitive
BPVYCXMGJPKOTQ-UHFFFAOYSA-N
2-[3-(trifluoromethyl)phenyl]ethanamine
104223
98%

Chemical Identifiers

52516-30-0
189.181
BPVYCXMGJPKOTQ-UHFFFAOYSA-N
104223
C1=CC(=CC(=C1)C(F)(F)F)CCN
C9H10F3N
MFCD00040756
2-3-trifluoromethyl phenyl ethanamine, 3-trifluoromethyl phenethylamine, 2-3-trifluoromethylphenyl ethylamine, 2-3-trifluoromethyl phenyl ethylamine, phenethylamine, m-trifluoromethyl, m-trifluoromethylphenethylamine, m-trifluoromethyl phenethylamine, benzeneethanamine, 3-trifluoromethyl, 2-3-trifluoromethylphenyl-ethylamine
2-[3-(trifluoromethyl)phenyl]ethanamine
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.

EINECSNumber : 257-988-3

RTECSNumber : SI2525000

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only