missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-[4-(1,3,4-Oxadiazol-2-yl)phenoxy]acetamidoxime, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H50673MD

 View more versions of this product

Catalog No. AAH50673MD


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-[4-(1,3,4-Oxadiazol-2-yl)phenoxy]acetamidoxime
C10H10N4O3
250 mg
RNDSGEINHPCNQK-UHFFFAOYSA-N
N'-hydroxy-2-[4-(1,3,4-oxadiazol-2-yl)phenoxy]ethanimidamide
57376339
97%
258521-20-9
MFCD01764763
2-4-1,3,4-oxadiazol-2-yl phenoxy acetamidoxime, 1-hydroxyimino-2-4-1,3,4-oxadiazol-2-yl phenoxy ethylamine
NC(COC1=CC=C(C=C1)C1=NN=CO1)=NO
234.22
234.22

Chemical Identifiers

258521-20-9
234.22
RNDSGEINHPCNQK-UHFFFAOYSA-N
57376339
NC(COC1=CC=C(C=C1)C1=NN=CO1)=NO
C10H10N4O3
MFCD01764763
2-4-1,3,4-oxadiazol-2-yl phenoxy acetamidoxime, 1-hydroxyimino-2-4-1,3,4-oxadiazol-2-yl phenoxy ethylamine
N'-hydroxy-2-[4-(1,3,4-oxadiazol-2-yl)phenoxy]ethanimidamide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only