missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-[4-(1,3,4-Oxadiazol-2-yl)phenoxy]acetamidoxime, 97%, Thermo Scientific Chemicals

$114.88 - $325.05

Chemical Identifiers

CAS 258521-20-9
Molecular Formula C10H10N4O3
Molecular Weight (g/mol) 234.22
MDL Number MFCD01764763
InChI Key RNDSGEINHPCNQK-UHFFFAOYSA-N
Synonym 2-4-1,3,4-oxadiazol-2-yl phenoxy acetamidoxime, 1-hydroxyimino-2-4-1,3,4-oxadiazol-2-yl phenoxy ethylamine
PubChem CID 57376339
IUPAC Name N'-hydroxy-2-[4-(1,3,4-oxadiazol-2-yl)phenoxy]ethanimidamide
SMILES NC(COC1=CC=C(C=C1)C1=NN=CO1)=NO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH50673MD
View Documents
Thermo Scientific Chemicals
H50673MD
250 mg N/A
 
AAH5067303
View Documents
Thermo Scientific Chemicals
H5067303
1 g N/A
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

258521-20-9
234.22
RNDSGEINHPCNQK-UHFFFAOYSA-N
57376339
NC(COC1=CC=C(C=C1)C1=NN=CO1)=NO
C10H10N4O3
MFCD01764763
2-4-1,3,4-oxadiazol-2-yl phenoxy acetamidoxime, 1-hydroxyimino-2-4-1,3,4-oxadiazol-2-yl phenoxy ethylamine
N'-hydroxy-2-[4-(1,3,4-oxadiazol-2-yl)phenoxy]ethanimidamide

Specifications

258521-20-9
MFCD01764763
2-4-1,3,4-oxadiazol-2-yl phenoxy acetamidoxime, 1-hydroxyimino-2-4-1,3,4-oxadiazol-2-yl phenoxy ethylamine
NC(COC1=CC=C(C=C1)C1=NN=CO1)=NO
234.22
234.22
2-[4-(1,3,4-Oxadiazol-2-yl)phenoxy]acetamidoxime
C10H10N4O3
250 mg
RNDSGEINHPCNQK-UHFFFAOYSA-N
N'-hydroxy-2-[4-(1,3,4-oxadiazol-2-yl)phenoxy]ethanimidamide
57376339
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only