Learn More
2,4,4-Trimethyl-2-pentene, 98%, Thermo Scientific Chemicals
$120.74 - $120.74
Chemical Identifiers
CAS | 107-40-4 |
---|---|
Molecular Formula | C8H16 |
Molecular Weight (g/mol) | 112.21 |
MDL Number | MFCD00008902 |
InChI Key | LAAVYEUJEMRIGF-UHFFFAOYSA-N |
Synonym | 2,4,4-trimethyl-2-pentene, 2-pentene, 2,4,4-trimethyl, 2,2,4-trimethyl-3-pentene, 2,4,4-trimethylpentene-2, unii-548r0du21i, beta-diisobutylene, propene, 1-tert-butyl-2-methyl, acmc-1cujo, dsstox_cid_6766, dsstox_rid_78208 |
PubChem CID | 7869 |
IUPAC Name | 2,4,4-trimethylpent-2-ene |
SMILES | CC(=CC(C)(C)C)C |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC162630050
|
Thermo Scientific Chemicals
162630050 |
5 mL | Glass bottle |
Each for $120.74
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
107-40-4 | |
112.21 | |
LAAVYEUJEMRIGF-UHFFFAOYSA-N | |
7869 | |
CC(=CC(C)(C)C)C |
C8H16 | |
MFCD00008902 | |
2,4,4-trimethyl-2-pentene, 2-pentene, 2,4,4-trimethyl, 2,2,4-trimethyl-3-pentene, 2,4,4-trimethylpentene-2, unii-548r0du21i, beta-diisobutylene, propene, 1-tert-butyl-2-methyl, acmc-1cujo, dsstox_cid_6766, dsstox_rid_78208 | |
2,4,4-trimethylpent-2-ene |
Specifications
107-40-4 | |
100.0 | |
Colorless to Yellow | |
104.0°C | |
Authentic | |
Glass bottle | |
1.4150 to 1.4170 | |
MFCD00008902 | |
01, III, 848 | |
2,4,4-trimethyl-2-pentene, 2-pentene, 2,4,4-trimethyl, 2,2,4-trimethyl-3-pentene, 2,4,4-trimethylpentene-2, unii-548r0du21i, beta-diisobutylene, propene, 1-tert-butyl-2-methyl, acmc-1cujo, dsstox_cid_6766, dsstox_rid_78208 | |
CC(=CC(C)(C)C)C | |
112.21 | |
112.21 | |
Liquid |
97.5 | |
-106.0°C | |
0.7200g/mL | |
2°C | |
98% | |
C8H16 | |
(CH3)3CCH=C(CH3)2 | |
5 mL | |
0.72 | |
LAAVYEUJEMRIGF-UHFFFAOYSA-N | |
2,4,4-trimethylpent-2-ene | |
7869 | |
98% | |
2, 4, 4-Trimethyl-2-pentene, 98% |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapour.
May be fatal if swallowed and enters airways.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Toxic to aquatic life with lon
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or ha
GHS Signal Word: Danger
EINECSNumber : 203-488-5
TSCA : TSCA
RUO – Research Use Only