missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,4,6-Tribromoaniline, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1245322

 View more versions of this product

Catalog No. AAA1245322


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2,4,6-Tribromoaniline
118°C to 122°C
300°C
MFCD00007634
2209258
benzenamine, 2,4,6-tribromo, s-tribromoaniline, 2,4,6-tribromophenylamine, 2,4,6-tribromobenzenamine, sym-tribromoaniline, aniline tribromide, tribromoaniline, usaf do-43, aniline, 2,4,6-tribromo, 2,4,6-tribomoaniline
NC1=C(Br)C=C(Br)C=C1Br
329.82
329.83
147-82-0
2.35
C6H4Br3N
100 g
14,9610
GVPODVKBTHCGFU-UHFFFAOYSA-N
2,4,6-tribromoaniline
8986
98%

Chemical Identifiers

147-82-0
329.82
GVPODVKBTHCGFU-UHFFFAOYSA-N
8986
NC1=C(Br)C=C(Br)C=C1Br
C6H4Br3N
MFCD00007634
benzenamine, 2,4,6-tribromo, s-tribromoaniline, 2,4,6-tribromophenylamine, 2,4,6-tribromobenzenamine, sym-tribromoaniline, aniline tribromide, tribromoaniline, usaf do-43, aniline, 2,4,6-tribromo, 2,4,6-tribomoaniline
2,4,6-tribromoaniline
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P260-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P314-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335-H373

DOTInformation : Hazard Class: 6.1; Packaging Group: III

EINECSNumber : 205-700-1

RTECSNumber : BZ0175000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only