Learn More
2,4,6-Tribromoaniline, 98%, Thermo Scientific Chemicals
$79.99 - $334.19
Chemical Identifiers
CAS | 147-82-0 |
---|---|
Molecular Formula | C6H4Br3N |
Molecular Weight (g/mol) | 329.82 |
MDL Number | MFCD00007634 |
InChI Key | GVPODVKBTHCGFU-UHFFFAOYSA-N |
Synonym | benzenamine, 2,4,6-tribromo, s-tribromoaniline, 2,4,6-tribromophenylamine, 2,4,6-tribromobenzenamine, sym-tribromoaniline, aniline tribromide, tribromoaniline, usaf do-43, aniline, 2,4,6-tribromo, 2,4,6-tribomoaniline |
PubChem CID | 8986 |
IUPAC Name | 2,4,6-tribromoaniline |
SMILES | NC1=C(Br)C=C(Br)C=C1Br |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1245322
|
Thermo Scientific Chemicals
A1245322 |
100 g |
Each for $79.99
|
|
|||||
AAA1245336
|
Thermo Scientific Chemicals
A1245336 |
500 g |
Each for $334.19
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
147-82-0 | |
329.82 | |
GVPODVKBTHCGFU-UHFFFAOYSA-N | |
8986 | |
NC1=C(Br)C=C(Br)C=C1Br |
C6H4Br3N | |
MFCD00007634 | |
benzenamine, 2,4,6-tribromo, s-tribromoaniline, 2,4,6-tribromophenylamine, 2,4,6-tribromobenzenamine, sym-tribromoaniline, aniline tribromide, tribromoaniline, usaf do-43, aniline, 2,4,6-tribromo, 2,4,6-tribomoaniline | |
2,4,6-tribromoaniline |
Specifications
147-82-0 | |
2.35 | |
C6H4Br3N | |
100 g | |
14,9610 | |
GVPODVKBTHCGFU-UHFFFAOYSA-N | |
2,4,6-tribromoaniline | |
8986 | |
98% |
118°C to 122°C | |
300°C | |
MFCD00007634 | |
2209258 | |
benzenamine, 2,4,6-tribromo, s-tribromoaniline, 2,4,6-tribromophenylamine, 2,4,6-tribromobenzenamine, sym-tribromoaniline, aniline tribromide, tribromoaniline, usaf do-43, aniline, 2,4,6-tribromo, 2,4,6-tribomoaniline | |
NC1=C(Br)C=C(Br)C=C1Br | |
329.82 | |
329.83 | |
2,4,6-Tribromoaniline |
Safety and Handling
GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P260-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P314-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335-H373
DOTInformation : Hazard Class: 6.1; Packaging Group: III
EINECSNumber : 205-700-1
RTECSNumber : BZ0175000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only